Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Molecular Biology > Automation > GenElute HP 96 Well Miniprep Kit
Automation
GenElute HP 96 Well Miniprep Kit
GenElute HP 96 Well Plasmid Miniprep Kit Image Simple, rapid, high-throughput purification of transfection quality plasmid DNA in less than 50 minutes
The GenElute HP 96 Well Plasmid Miniprep Kit has been developed for use as a high-throughput method for the purification of plasmid DNA from recombinant E. coli cultures. This rapid and cost-effective system allows for the purification of up to 10 mg of high copy plasmid DNA per well. Purified DNA is of high quality and can be used in a variety of molecular biology applications including transfection, cloning, restriction digestion, in-vitro transcription and translation, and sequencing. Designed to complement Sigma’s MISSION® shRNA product line, the GenElute HP 96-Well Plasmid Miniprep Kit is optimized for the most demanding downstream transfection protocols. The result is purified plasmid DNA that can generate best-in-class transfection grade plasmid that delivers the highest viral titers of any 96-well plasmid kit on the market.
The GenElute HP 96 Well Plasmid Miniprep Kit is compatible with liquid handling instrumentation. An automated method has been developed for the Biomek® FX from Beckman Coulter. The protocol is simple and rapid, resulting in a processing time of less than 50 minutes.
Automation Resources
The following automation resources are available for download:
Applications
Purified plasmid DNA is suitable for a variety of applications including:
  • Transfection
  • Cloning
  • In vitro transcription and translation
  • Restriction enzyme digestion
  • Sequencing
Features & Benefits
Transfection-Grade Quality Best-in-class production of viral titers from purified MISSION shRNA plasmid vectors.
High Yields Up to 10 µg of high copy plasmid DNA per well.
Rapid Procedure 96 samples can be processed in less than 50 minutes.
Versatile Generates purified plasmid DNA for a wide variety of downstream applications.
GenElute HP 96 Plasmid Miniprep Kits
Product #  Product Name Package Size
NA9604 GenElute HP 96 Plasmid Miniprep Kits 4 x 96
GenElute is a trademark of Sigma-Aldrich, Inc.
MISSION is a trademark of Sigma-Aldrich, Inc.
Biomek FX is a registered trademark of Beckman Coulter, Inc.
back to top
back to Automation