High Efficiency Competent Cells for your Sensitive Cloning Applications!
Sigma-Aldrich offers a wide range of chemically competent cells as well as Thunderbolt™ GC10™ electrocompetent cells. Our competent cells are offered at the highest transformation efficiencies for sensitive cloning applications. In addition, we offer a variety of strains to meet your specific needs for protein expression and plasmid propagation.
Features and Benefits
  • Convenient Unipack formats for one time use
  • Strains comparable to DH5α™ and DH10B™
  • High efficiencies
  • T1 phage resistance
Competent Cell Compendium
Cloning Strains
Expression Strains
Competent Cell Selection Table
The Competent Cell Compendium: Tips and Tools for Successful Transformations" Now Available
Download your copy today!
(765 Kb pdf)
or
Launch the eBook today!
Other Resources:
     Watch the new technical presentation "Everything you Need to Know about Competent Cells!"
     Calculating Transformation Efficiences
     Genotypes, Phenotypes and Markers - Definitions
     E. coli Markers
     Strains of E. coli
Cloning Strains
Features and Benefits
  • F' episome for single strand rescue
  • Strains for cloning methylated DNA
  • Blue/white selection
Product # 
Product Name
Size
G3169
GC5™ Competent Cells, Uni-pack
10 x 50 µl
20 x 50 µl
G3044
GC5™ Competent Cells, Standard Aliquots
5 x 200 µl
G7419
GC5™ Competent Cells, 96-well
1 EA
4 EA
G2919
GC10™ Competent Cells, Uni-Pack
10 x 50 µl
20 x 50 µl
G2794
GC10™ Competent Cells, Standard Aliquots
5 x 200 µl
T7699
Thunderbolt™ GC10™ ElectroCompetent Cells
5 x 80 µl
5 x 100 µl
J3895
JM109 Competent Cells, Uni-pack
10 x 50 µl
H3788
HB101 Competent Cells, Uni-pack
10 x 50 µl
back to top
Expression Strains
Features and Benefits
  • T1 phage resistance
  • Strains for T7 systems
  • Ion and omp T protease deficient
Product # 
Product Name
Size
B2685
BL21-T1R Competent Cells, Uni-pack
10 x 50 µl
B2935
BL21 (DE3)-T1R Competent Cells, Uni-pack
10 x 50 µl
B3310
BL21 (DE3) pLysS-T1R Competent Cells, Uni-pack
10 x 50 µl
B3435
BL21 (DE3) pLysE-T1R Competent Cells, Uni-pack
10 x 50 µl
back to top
Competent Cell Strain Selection Table:
Feature GC5™ GC10™ Thunderbolt™ GC10™ JM109 HB101 BL21 BL21 (DE3) BL21 (DE3) pLysS BL21 (DE3) pLysE
Comparable to DH5a
X
Comparable to NovaBlue and XL1 Blue
X
Comparable to DH10B™
X
X
Strain Background
K12
K12
K12
K12
K12xB
B
B
B
B
Transformation Efficiency (cfu/ug pUC plasmid DNA)
109
109
1010
108
108
106
107
106
106
Cloning Strain
X
X
X
X
X
Expression Strain
X
X
X
X
Blue/White Selection (with appropriate construct)
X
X
X
X
T1 bacteriophage resistant
X
X
X
X
X
X
X
T7 promoter systems
X
X
X
F' episome for single strand rescue
X
Ideal for cloning methylated DNA
X
X
X
lacI q (higher level of lac repressor)
X
GC5™, GC10™, and Thunderbolt™ are trademarks of GeneChoice ®, Inc.
DH5α™ and DH10B™ are trademarks of Invitrogen Corp.
back to top
back to Cloning & Expression
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page