|
Cloning & Expression
BioPORTER® QuikEase™ Protein Delivery Kit
|
Product No. BPQ24 suitable for 24 reactions Product No. BPQ96 suitable for 96 reactions
When you need to deliver biologically active proteins into living cells quickly and effectively, at a reduced cost — BioPORTER Protein Delivery Kit is the ideal option.
The BioPORTER reagent reacts quickly and interacts non-covalently with the protein, peptide or other molecules, creating a protective vehicle for immediate delivery into cells. Various molecules (fluorescent-antibody, high and low molecular weight dextran sulfate, caspase 3 and 8, phycoerythrin-BSA, and others) have been successfully delivered into the cytoplasm of a variety of adherent and suspension cells with the BioPORTER reagent. Furthermore, apoptotic proteins like granzyme B, caspase 3 and caspase 8 that were delivered into cells remained functional and induced apoptosis in cultured cells.
|
BioPORTER is a valuable tool for a variety of studies, including:
- Intercellular signaling
- Cell cycle regulation
- Apoptosis control
- Oncogenesis
- Transcripton regulation
|
|
Features & Benefits
- Easy to Use
- More economical than microinjections or electroporation
- Reaches optimum levels after four hours of incubation
- Successfully delivers various molecules such as; fluorescent antibody, high and low molecular weight dextran sulfate, caspase 3 and 8, phycoerythrin-BSA, and others.
- Delivers effectively into a variety of adherent and suspension cells
- Apoptopic proteins delivered with BioPORTER have been found to remain functional and induce apoptosis in cultured cells.
back to top Back to Cell Signaling Home