For Bacterial, Mammalian and Insect Cell Systems
Featured are a variety of tags, promoters and elements for secretion, transient, stable and bicistronic expression.
- Our exclusive FLAG® and 3xFLAG® tags are provided in N-terminal, C-terminal, and dual tag configurations in bacterial, mammalian and insect vectors for high quality detection and purification using ANTI-FLAG® antibodies, resins, and 96-well plates.
- Unique BICEP™ vectors contain an optimized EMCV IRES for high levels of bicistronic expression in mammalian cells for optimal maintenance of stable cell lines and multi-gene expression.
|
|
- The new MAT™ (Metal Affinity Tag) has been incorporated in many of our popular bacterial, mammalian, and insect vectors for high quality downstream purification using HIS-Select™ affinity gels.
- Sigma's pPolh transfer vectors are the first to offer the well-known FLAG tag for expression of FLAG fusions in insect cell systems.
Bacterial Expression Vectors
Mammalian Expression Vectors
Insect Expression System
Vectors by Affinity Tag
back to top back to Cloning & Expression
|