Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Molecular Biology > Cloning & Expression >  Expression Vectors > Mammalian FLAG Expression Kits
Cloning & Expression
Mammalian FLAG Expression Kits
Mammalian Amino-terminal FLAG® Transient Expression Kit (FL-MA)
The Mammalian Amino-Terminal FLAG® Transient Expression Kit provides all of the specialized FLAG components needed to perform expression of a recombinant N-terminal FLAG fusion in mammalian cells. The pFLAG-CMV™-1 (E 7273) and pFLAG-CMV™-2 (E 7398) Expression Vectors allow transient expression of secreted or cytoplasmically expressed fusion proteins. Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M2 monoclonal antibody
  • ANTI-FLAG® M5 monoclonal antibody
  • C-CMV-24 sequencing primer
  • N-CMV-30 sequencing primer
  • FLAG® peptide
  • Met-FLAG-BAP™ control protein
  • pFLAG-CMV™-1-BAP control plasmid
  • pFLAG-CMV™-2-BAP control plasmid
  • pFLAG-CMV™-2 expression vector (no leader)
  • pFLAG-CMV™-1 expression vector (preprotrypsin leader)
 pFLAG  CMV 1
 pFLAG CMV 2
Product Code Description Size
FL-MA Mammalian Amino-Terminal FLAG® Transient Expression Kit 1 kit
back to top
Mammalian Amino-terminal FLAG® Stable Expression Kit (FL-MA-S)
The Mammalian Amino-Terminal FLAG Stable Expression Kit provides the specialized FLAG components needed for expression of a recombinant N-terminal FLAG fusion in mammalian cells. The pFLAG-CMV™-3 (E 8770) and pFLAG-CMV™-4 (E 1775) Expression Vectors allow transient or stable expression of secreted or cytoplasmically expressed fusion proteins. Stable expression is accomplished by neomycin selection (G 418). Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M2 monoclonal antibody
  • ANTI-FLAG® M5 monoclonal antibody
  • C-CMV-24 sequencing primer
  • N-CMV-30 sequencing primer
  • FLAG® peptide
  • Met-FLAG-BAP™ control protein
  • pFLAG-CMV™-3-BAP control plasmid
  • pFLAG-CMV™-4-BAP control plasmid
  • pFLAG-CMV™-4 expression vector (no leader)
  • pFLAG-CMV™-3 expression vector (preprotrypsin leader)
 pFLAG CMV 3
pFLAG CMV 4 
Product Code Description Size
FL-MA-S Mammalian Amino-terminal FLAG® Stable Expression Kit 1 kit
back to top
Mammalian Carboxy-terminal FLAG® Transient Expression Kit (FL-MC)
The Mammalian Carboxy-Terminal FLAG® Transient Expression Kit provides the specialized FLAG components needed for transient expression of a recombinant C-terminal FLAG fusion in mammalian cells. The pFLAG-CMV™-5a,b,c Expression Vectors (E 3762) make all reading frames of the MCS available at each restriction site for cloning of an open reading frame. All three vectors lack a translational start signal, so the start signal and any secretion leaders must be inserted along with the gene of interest. Also provided in the kit are controls for expression, detection, and affinity purification.
Components
  • ANTI-FLAG® M2 affinity gel
  • ANTI-FLAG® M2 monoclonal antibody
  • C-CMV-24 sequencing primer
  • N-CMV-30 sequencing primer
  • FLAG® peptide
  • pFLAG-CMV™-5b–BAP control plasmid
  • pFLAG-CMV™-5a expression vector
  • pFLAG-CMV™-5b expression vector
  • pFLAG-CMV™-5c expression vector
  • C-terminal FLAG–BAP™ control protein
 pFLAG CMV 5abc
Product Code Description Size
FL-MC Mammalian Carboxy-Terminal FLAG® Transient Expression Kit 1 kit
back to top
back to Expression Vectors