|
DNA & RNA Purification Kits
Extract-N-Amp™ Blood PCR Kits
|
|
|
Description
The Extract-N-Amp Blood PCR kits contain all of the reagents necessary to rapidly extract genomic DNA from whole blood and amplify targets of interest by PCR (Figure 1).
- Novel – single-step extraction of genomic DNA to PCR
- Fast – tissue to PCR in 15 minutes
- Convenient – no long enzymatic digestions
The Extract-N-Amp Blood PCR Kit offers a novel extraction system that eliminates the need for DNA purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The product includes a specially formulated hot start PCR ReadyMix™ for amplification directly from the extract. Extract-N-Amp products are Sigma Advanced Technology certified.
Procedure
Genomic DNA is extracted from 10 µl of whole blood by adding Extraction Solution and incubating for 5 minutes at room temperature. After adding Neutralization Solution, this crude blood extract is stable for at least 6 months at 4 °C and 3 months at 37 °C (Figure 2) and can be added directly to the optimized PCR mix supplied.
Application
Perfect for cloning and automated sequencing protocols.
Beneficial Features
| Starting Material |
10 µl of blood |
| Speed |
Extraction of genomic DNA for PCR in 8 minutes |
| Scalable |
Liquid format fits any configuration (single tube, 96-well, etc.) (Figure 1) |
| Specific |
Hot Start antibody for highly specific PCR amplification of genomic DNA |
| Safe |
No phenol/chloroform or other hazardous materials |
Pick the formulation that is right for you
The Extract-N-Amp Tissue PCR Kits include a specially formulated hot start PCR ReadyMix for amplification directly from the extract.
The PCR ReadyMix comes in two formulations:
- REDExtract-N-Amp PCR Ready Mix – contains an inert dye that acts as a tracking dye and allows for convenient loading of PCR reactions onto agarose gels for analysis.
- Extract-N-Amp PCR Ready Mix – no dye contained.
Ordering Information
Extract-N-Amp Blood PCR Kits
| Product No. |
Name |
Number of Extractions |
Number of Amplifications |
Technical Manual .pdf |
| XNAB2 |
Extract-N-Amp Blood PCR Kit |
100 |
100 |
X |
| XNAB2R |
Extract-N-Amp Blood PCR Kit |
1000 |
1000 |
X |
| XNAB2RE |
Extract-N-Amp Blood PCR Kit |
1000 |
5000 |
X |
REDExtract-N-Amp Blood PCR Kits
| Product No. |
Name |
Number of Extractions |
Number of Amplifications |
Technical Manual .pdf |
| XNABS |
REDExtract-N-Amp Blood PCR Kit (contains REDTaq) |
10 |
10 |
X |
| XNAB |
REDExtract-N-Amp Blood PCR Kit (contains REDTaq) |
100 |
100 |
X |
| XNABE |
REDExtract-N-Amp Blood PCR Kit (contains REDTaq) |
100 |
500 |
X |
| XNABR |
REDExtract-N-Amp Blood PCR Kit (contains REDTaq) |
1000 |
1000 |
X |
| XNABRE |
REDExtract-N-Amp Blood PCR Kit (contains REDTaq) |
1000 |
5000 |
X |
Kit Contents
Extract-N-Amp Blood PCR Kit
- Extraction Solution
- Neutralization Solution
- REDExtract-N-Amp and Extract-N-Amp PCR Ready Mix
Sample Data
PCR analysis of genomic DNA isolated from blood using Sigma's Extract-N-Amp Blood PCR Kit
| Figure 1. Extract-N-Amp Blood PCR Kit used to isolate genomic DNA from fresh, 7 day old & frozen blood. DNA was extracted and neutralized from 10 µl of whole blood in 5 minutes at room temperature using the REDExtract-N-Amp Blood PCR kit. The PCR products were then generated using the specially formulated Hot Start PCR mix included in the kit. PCR products generated are 1.8 kb for carnitine palmitoyltransferase II, 1.3 kb for a mitochondrial DNA control region, 547 bp for human surfactant protein B and 320 bp for the 5' untranslated region of human major histocompatibility complex class II. |
 |
Stability of Extract-N-Amp Blood Extracts
| Figure 2. Blood was drawn from 2 human volunteers into vacutainer tubes containing EDTA. Extractions were performed in duplicate providing 4 samples total. Samples were stored at 37 °C and removed at various time intervals for testing. Stability was determined by monitoring yield from quanitative PCR using an ABI 7700 instrument. The DNA standards used for the quanitative PCR were generated from the same blood drawn as the test samples. The DNA for the standards was purified using the GenElute Blood Genomic DNA Kit (NA2000). The PCR products were generated using primers for a 547 bp product for human surfactant protein B (SPB; Lin & Floros, 2000, BioTechniques, 29:460-466). The results clearly show no loss of amplification of the SPB PCR product even after storage at 37 °C for 6 months. |
 |
return to top