Sequence Determination
Sequence was resolved on an ABI 310 from a purified, 645 bp corn leaf PCR product. The PCR product was purified with the GenElute PCR Clean-Up Kit (Product Code NA1020). The DNA extraction and PCR were performed using Sigma's Extract-N-Amp Plant PCR Kit. The sequence was obtained using the same primers as for the original PCR.
Close Window