Sigma-Aldrich
Life Science > Molecular Biology > DNA & RNA Purification > Extract-N-Amp™ Reagents
DNA & RNA Purification
Extract-N-Amp™ Reagents
Get more from your Extract-N-Amp PCR Kit!
Sigma® offers Extract-N-Amp supplementary reagents to use in conjunction with our Extract-N-Amp kits. The PCR ReadyMixes allow increased amplifications from the Extract-N-Amp PCR Kits, and accommodate dilution experiments prior to PCR amplification.
Sigma Extract-N-Amp PCR Kits offer an innovative extraction system that prepares genomic DNA for amplification by PCR or qPCR in as little as 8 minutes. The optimized kits include a specially formulated hot start PCR ReadyMix™ for amplification directly from an aliquot of the stable extract. Suitable for most downstream applications, our Extract-N-Amp kits provide the perfect solution for genomic DNA analysis.
Rapid Genotyping of Mouse Tissue Using Sigma's Extract-N-Amp™ Tissue PCR Kit
Watch the video protocol
Rapid Genotyping of Mouse Tissue Using Sigma's Extract-N-Amp Tissue PCR Kit
Video brought to you by JoVE.com
Re-amplification mixes for Extract-N-Amp Tissue and Plant Kits
Product No. Name Package Size Add to Cart
E3004 Extract-N-Amp PCR ReadyMix 1.2 ml
12 ml
125 ml
R4775 REDExtract-N-Amp™ PCR ReadyMix 1.2 ml
12 ml
125 ml
S4320 SYBR® Green Extract-N-Amp PCR ReadyMix 1.2 ml
12 ml
125 ml
Re-amplification mixes for Extract-N-Amp Blood Kits
Product No. Name Package Size Add to Cart
P8115 Extract-N-Amp PCR ReadyMix for Blood 12 ml
P8240 REDExtract-N-Amp PCR ReadyMix for Blood 12 ml
Supplementary PCR diluent for Extract-N-Amp Tissue, Plant and Seed Kits
Product No. Name Package
Size
Add to Cart
E8155 Extract-N-Amp PCR Diluent 100 ml
E8155 Extract-N-Amp PCR Diluent 500 ml
back to top
DNA & RNA Purification home page