Life Science > Molecular Biology > DNA & RNA Purification > GenElute™ Agarose Spin Columns
DNA & RNA Purification
GenElute™ Agarose Spin Columns
Description: GenElute™ Agarose Spin Columns isolate DNA by centrifugation and filtration.
Procedure: A slice of agarose gel containing the DNA of interest is excised, placed into a spin column, and spun at maximum speed in a microcentrifuge. Agarose is retained on a filter in the spin column. Soluble molecules, including DNA, pass through the filter and collect in a microcentrifuge tube (Figure 1).
Application: DNA can be used in ligation, restriction enzyme digestion, cloning and PCR
Purification Chart
Beneficial Features
Starting Amount
< 200 mg gel slice
Yield
Recovers up to 40 to 45% of DNA fragments from 100 bp to 10 kb
Speed
One-step, 10 minute protocol
Flexible
DNA concentrated in 50 µL to 200 µL of buffer depending on size of gel slice
Return to Top
Order Information
Product Code Product Name Purifications
per kit
Technical Manual .pdf
5-6500 GenElute Agarose Spin Columns 70 X
Return to Top
Sample Data
Recovery of 100-600 bp and 2-23 kb DNA fragments purified using GenElute Agarose Spin Columns.
Recovery of DNA Fragments
Recovery of DNA Fragments
Left Panel: 1 µg of 100 bp DNA ladder was electrophoresed on a 2.0% agarose gel. 100, 200, 300, 400, 500, and 600 bp fragments were excised from the gel, and purified using GenElute Agarose Spin Columns as above. Samples were loaded and analyzed on a 2.0% agarose gel.
Right Panel: Lambda Hind III markers were pre-heated at 65°C and electrophoresed on a 2.0% agarose gel. The 2.0, 4.4, 6.6, 9.4, and 23 kb fragments were excised and purified using GenElute Agarose Spin Columns. Samples were loaded and analyzed on a 2.0% agarose gel.
Return to Top