|
DNA & RNA Purification
Genomic DNA
|
|
We provide Genomic DNA Purification kits and reagents for the purification of mammalian genomic DNA, plant genomic DNA, blood genomic DNA, bacterial genomic DNA. The Genomic DNA Purification kits are useful in PCR, restriction endonuclease digestions, cloning, southern blots and sequencing.
Click on the product for more information
| Product |
Yield |
Speed |
Starting Material |
Product Description |
Technology |
Applications |
| GenElute™ Mammalian Genomic DNA Miniprep Kit |
Up to 30 µg, varies depending on starting material |
< 40 minutes |
Up to 25 µg from 2 x 106 Cells or 30 µg from 25 mg of Tissue |
Purification of genomic DNA from a variety of mammalian sources |
Advanced silica bind & elute in a spin column format |
PCR, restriction endonuclease digestions, cloning and southern blots |
| GenElute™ Plant Genomic DNA Miniprep Kit |
Up to 11.5 µg, varies depending on plant species |
< 40 minutes |
100 mg of Plant Tissue |
Purification of genomic DNA from a variety of plant species |
Advanced silica bind & elute in a spin column format |
PCR, restriction endonuclease digestions, cloning and southern blots |
| GenElute™ Blood Genomic DNA Kit |
Up to 10 µg |
< 40 minutes |
Up to 500 µl of Blood |
Purification of genomic DNA from fresh or aged whole blood |
Advanced silica bind & elute in a spin column format |
PCR, restriction endonuclease digestions, cloning, southern blots, and sequencing |
| GenElute™ Bacterial Genomic DNA Kit |
Up to 20 µg |
<75 minutes Gram -
<120 minutes Gram + |
Up to 1.5 ml of culture |
Isolation of pure genomic DNA from a variety of cultured bacteria |
Advanced silica bind & elute in a spin column format |
PCR, restriction endonuclease digestions, cloning, southern blots, and sequencing |