Sigma-Aldrich
Life Science > Molecular Biology > DNA & RNA Purification > GenElute™ HP Plasmid Miniprep Kits
DNA & RNA Purification
GenElute™ HP Plasmid Miniprep Kits
Description: The GenElute™ HP Plasmid Miniprep Kits offer purification of high-quality plasmid DNA in less than 30 minutes. These kits provide the convenience of binding columns that can be used with a vacuum or spin purification format. In addition, no alcohol precipitation is required.
  • Sophisticated – Sigma's GenElute HP technology ensures improved performance
  • Fast – Complete multiple preps in less than 30 minutes
  • High Yields – Up to 25 µg of high-quality plasmid DNA
  • Economic – lower cost + higher yields = significant savings
Procedure: An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the plasmid DNA onto silica in the presence of high salts. Contaminants are removed by a vacuum or spin wash step. Finally, the bound plasmid DNA is eluted in water or Tris-EDTA buffer.
Application: High-quality DNA is ready for immediate use for the most demanding applications, including transfection, restriction digestion, ligation, automated sequencing and PCR.
Resuspend overnight culture and lyse cells.
Neutralize and clear lysate.
Prepare binding column.
Load cleared lysate onto binding column and wash.
Elute plasmid DNA.
HP-Miniprep-protocol-2
Return to Top
Beneficial Features:
Yield Up to 25 µg of high-copy plasmid DNA
Speed Isolate high-quality DNA in less than 30 minutes
Flexibility Offers the convenience of a vacuum or spin format
Economic Lower cost per prep plus higher yields per prep equals double savings
Ordering Information
Product Code
Product Name
Preparations per Kit
Technical Manual
NA0150S
GenElute™ HP Plasmid Miniprep Kit
10
Technical_Bulletin_icon
NA0150
GenElute™ HP Plasmid Miniprep Kit
70
NA0160
GenElute™ HP Plasmid Miniprep Kit
350
Return to Top
Sample Data
Superior yield of plasmid DNA in less time.
Average Yield per Prep
Click on image to enlarge
Average Time per Prep
Click on image to enlarge
Figure 1 and Figure 2. Comparison of plasmid yield and time/prep for the different purification systems used for isolating plasmid pCMV1-Sport-βgal. The table on the left depicts the average yield per preparation obtained by the Sigma GenElute HP Plasmid Miniprep as opposed to kits from Supplier Q and Supplier P. The figure on the right depicts the average time per preparation of the GenElute HP Plasmid Miniprep as compared to kits from Supplier Q and Supplier P. All plasmid DNA was isolated following each manufacturers' recommended protocol.
Return to Top
High-quality plasmid DNA for Automated Sequencing.
Figure 3. Capillary DNA sequencing of plasmid DNA purified with the GenElute HP Plasmid Miniprep kit. pCMV-SPORT-βgal (7.9 kb) was prepared from 400 µl overnight culture of E. coli host strain DH5α with the GenElute HP Plasmid Miniprep Kit. A 5 µl sample of the preparation was submitted, with adjustment in DNA concentration, to SeqWright DNA Technology Service (Houston, TX) for sequencing witha T7 sequencing primer and ABI BigDye™ terminator v3.1 on an ABI Prism® 3730 DNA Analyzer. The Phred > 20 score was greater than 800 bases.
High Quality Plasmid DNA for Automated Sequencing
Click on image to enlarge
Return to Top