| Product |
Product No. |
Yield |
Speed |
Starting Material |
Product Description |
Technology |
Applications |
| GenElute™ 96 Well Blood Genomic DNA Purification Kit |
BLD9601 BLD9604 |
4 to 6 µg per well |
70 min/plate |
200 µL fresh blood, frozen blood or blood treated with EDTA, citrate, or heparin |
Rapid isolation of genomic DNA from whole blood |
Silica membrane with vacuum or spin format |
PCR, restriction digestion, cloning and southern blots |
| GenElute 96 Well Tissue Genomic DNA Purification Kit |
G1N9602 G1N9604 |
12 to 15 µg per well |
70 min/plate |
Less than 20 mg tissue or less than 10µL cultured cells or bacteris |
Rapid isolation of DNA from human and animal tissue, cultured cells and bacteria |
Silica membrane with vacuum or spin format |
PCR, restriction digestion, cloning and southern blots |
| GenElute 96 Well PCR Clean-up Kit |
PCR9601 PCR9604 |
75-95% recovery |
45 min/plate |
Less than 100 µL PCR reaction |
Complete removal of primers and primer-dimers |
Silica membrane with vacuum or spin format |
Capillary sequencing, Microarray analysis, ligation and restriction digestion |
| GenElute HP 96-Well Plasmid Miniprep Kit |
NA9604 |
Up to 10 µg |
45 min/plate |
1.3 mL bacterial culture per well |
Rapid purification of transfection grade plasmid |
Silica membrane with vacuum or spin format |
Transfections (robust cells, sequencing, restriction digestion, cloning and PCR |
| GenElute 96 Well Total RNA Purification Kit |
RTN9602 RTN9604 |
50 to 200 ng/µL |
70 min/plate |
Less than 107 cultured cells, less than 30 mg tissue or saliva |
Purification of total RNA from animal or himan cell cultures and tissue |
Silica membrane with vacuum or spin format |
RT-PCR, northern blots and microarray analysis |