Life Science > Molecular Biology > DNA & RNA Purification > Mammalian Genomic DNA Miniprep Kit
DNA & RNA Purification
Mammalian Genomic DNA Miniprep Kit
For purification of genomic DNA from a variety of mammalian sources
Description: The GenElute Mammalian Genomic DNA Purification Kit provides a simple and convenient way to isolate pure, high molecular weight DNA from a variety of mammalian sources (Figure 1 and Figure 2). These kits use a silica-based membrane, specially selected for genomic DNA purification, in a convenient spin column format.
Quick look: G1N10, G1N70, G1N350
Procedure: Mammalian cells and tissues are lysed with a chaotropic salt-containing buffer to ensure denaturation of macromolecules. DNA is bound to the spin column membrane and the remaining lysate is removed by centrifugation. A filtration column is used to remove cell debris. After washing to remove contaminants, the DNA is eluted with buffer into a collection tube. The purified DNA may be used in many applications such as sequencing, cloning, blotting, restriction digestion, ligation, and PCR.
Features and Benefits:
  • Typical DNA yields of 25 µg from 2 x 106 cultured cells or 30 µg from 25 mg of tissue (Figure 4)
  • Preparation time is only 20 minutes after lysis
  • Purified genomic DNA has A260/A280 ratios between 1.6 and 1.9
  • No need for mechanical homogenization
  • 40% more purification preps offered than other leading supplier
Storage: Room Temperature
R: 20/21/22 S: 26-36
Application: Purified DNA may be used in many applications such as sequencing, cloning, blotting, restriction digestion (Figure 3), ligation and PCR (Figure 4).
GenElute Mammalian Genomic Flow Chart
Save time with the
Genomic DNA Miniprep Kit!
Unlike most kits on the market,
the elution buffer in our Sigma kit does not have to be heated.
Return to Top
Sample Data
Table 1. Yields produced using GenElute Mammalian Genomic DNA Purification Kits (with RNase and Proteinase K treatment).
Material Amount Typical Yield
Jurkat Cells (human) 2 x 106 Cells 5 - 10 µg
HEK293 Cells (human) 2 x 106 Cells 10 - 20 µg
HeLa Cells (human) 2 x 106 Cells 15 - 25 µg
Mouse Pancreas Tissue
20 mg 10 - 25 µg
Mouse Spleen Tissue 10 mg 10 - 25 µg
Mouse Thymus Tissue 16 mg 10 - 25 µg
Mouse Lung Tissue 20 mg 5 - 15 µg
Mouse Brain Tissue 16 mg 5 - 15 µg
Mouse Kidney Tissue 20 mg 10 - 25 µg
Mouse Liver Tissue 25 mg 10 - 30 µg
Return to Top
Genomic DNA purified from mammalian tissues.
Figure 1. Genomic DNA purified from mammalian tissues. Purified genomic DNA, from the indicated mouse tissues, was isolated with the GenElute™ Mammalian Genomic DNA Miniprep Kit. Genomic DNA (200 ng/lane) was analyzed on a 0.8% agarose gel to illustrate yield and integrity. Markers are lambda DNA digested with Hind II. (Product No. D9780)
Genomic DNA purified from mammalian tissues
Genomic DNA purified from cells using GenElute Mammalian Genomic DNA Purification Kit.
Figure 2. Genomic DNA purified from mammalian cells. Purified genomic DNA was isolated with the GenElute Mammalian Genomic DNA Purification Kit from 2 x 106 cells of the sources indicated. The genomic DNA (200 ng/lane) was analyzed on a 0.8% agarose gel. Marker is a Lambda Hind III digest (0.5-23 kb).
Genomic DNA purified from mammalian cells
Return to Top
Genomic DNA purified with GenElute Mammalian Genomic DNA Purification Kit is suitable for restriction enzyme digestions.
Figure 3. Genomic DNA (200 ng) was digested with EcoR I (10 units in 20 µl at 37°C for 4 hours) followed by electrophoresis (150 ng/lane) on a 0.8% agarose gel. The undigested DNA was incubated under the same conditions, but without EcoR I. Marker is a Lambda Hind III digest (0.5-23 kb).
 Mammalian Restriction Enzyme
Return to Top
Yield Comparison.
Figure 4. Yields of genomic DNA compared with leading Supplier Q using 2 x 200 µl elutions.
GenElute Mammalian Genomic Yield
Order Information
Product Code Name Purifications per kit Technical Manual .pdf Short
Protocol.pdf
G1N10 GenElute Mammalian Genomic DNA Miniprep 10 X X
G1N70 GenElute Mammalian Genomic DNA Miniprep 70 X X
G1N350 GenElute Mammalian Genomic DNA Miniprep 350 X X
Return to Top