|
DNA & RNA Purification
GenElute™ Minus EtBr Spin Columns
|
Description: Description: GenElute™ Minus EtBr Spin Columns are based on the GenElute Agarose Spin Column but incorporate a membrane for the selective removal of ethidium bromide (Figure 1).
Procedure: The DNA band is excised from an agarose gel and loaded onto the spin column. The membranes, embedded within the column, retain the agarose and ethidium bromide while allowing DNA to selectively pass through the column into a centrifuge tube during centrifugation. Up to 95% of ethidium bromide is removed from DNA with a simple 10 minute procedure. GenElute Minus EtBr Spin Columns can recover DNA fragments from 100 bp to 10 kb with typical recoveries of 30 to 35%.
Application: The purified DNA can be used directly for ligation, Restriction Enzyme Digestion, Cloning, PCR and related manipulations without ethanol precipitation.
|
Beneficial Features
| Starting Amount |
<200 mg gel slice
|
| Yield |
Recovers up to 30 to 35% of DNA fragments from 100 bp to 10 kb
|
| Speed |
One-step, 10 minute protocol
|
| Flexible |
DNA concentrated in 50 µL to 200 µL of buffer depending on size of gel slice
|
Return to Top
Order Information
| Product Code |
Product Name |
Purifications per kit |
Technical Manual .pdf |
| 5-6501 |
GenElute Minus EtBr Spin Columns |
70 |
X |
Return to Top
Sample Data
Ethidium Bromide removal demonstrated over a UV light box
|
Figure 1. DNA was electrophoresed in agarose gels containing ethidium bromide (0.5 µg/ml). Bands were excised, applied to both the Minus EtBr and Agarose Spin Columns and centrifuged. The final eluate from each column was compared under a UV light box. The filter within the GenElute Minus EtBr Spin Column retained Ethidium Bromide.
Left Tube: GenEluteTM Agarose Spin Column
Right Tube: GenElute Minus EtBr Spin Column
|
Return to Top