Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Molecular Biology > DNA & RNA Purification > GenElute™ mRNA Miniprep Kits
DNA & RNA Purification
GenElute™ mRNA Miniprep Kits
Description
Features and Benefits
RNA Kits
Kit Contents
Preparation Time Comparison
RNA Yield
Specific mRNA Yield
mRNA—Suitable for Northern Blots
Description
Procedures such as cDNA synthesis, expression profiling, and others, require separation of mRNA from the vastly more abundant rRNA and tRNA. The GenElute mRNA Kits provide convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA, or directly from mammalian cells and tissues.
For direct mRNA preparation, cells or tissues are disrupted with SDS/proteinase K digestion to release RNA and eliminate RNases. Both kit types use oligo dT30 covalently linked to 1 µm polystyrene beads to capture polyadenylated mRNA by hybridization. The polystyrene beads remain suspended during hybridization, eliminating the need for mixing or rocking, as is common for cellulose or magnetic particles. Polystyrene was also chosen because oligo (dT) polystyrene beads yield cleaner mRNA with fewer stringent washing steps than does the more commonly used oligo (dT) cellulose (2 or 3 wash steps versus 10 or more). With the GenElute kits, mRNA-bead complexes are washed on a microcentrifuge spin filter, and eluted into 10 mM Tris-HCl, pH 7.5. mRNA prepared with either kit is suitable for a variety of downstream applications such as Northern Blotting (Figure 4), Expression Array or Chip Hybridizations, and cDNA Synthesis and Library Construction.
GenElute mRNA Miniprep Flow Chart
Protocol for GenElute mRNA Miniprep Kits
Kit Contents
Features and Benefits
Fast Prep time is 40 minutes for poly A+ mRNA from total RNA or 60 minutes directly from cells and tissues.
Convenient mRNA captured on polystyrene beads in 10 minutes, with no mixing or rocking. Beads washed on microcentrifuge spin filters.
Yield Higher than all competing products.
back to top
GenElute mRNA From Total RNA Kits
Product No.
Name
Purifications per Kit
Capacity
(µg Total RNA)
Technical Manual
Short Protocols
GenElute mRNA Miniprep Kit
10
5-500
GenElute mRNA Miniprep Kit
70
5-500
GenElute Direct mRNA Kits
Product No.
Name
Purifications per Kit
Capacity
Technical Manual
Short Protocols
GenElute Direct
mRNA Miniprep Kit
10
up to 107    up to 50mg
cells         tissues
GenElute Direct
mRNA Miniprep Kit
70
up to 107    up to 50mg
cells         tissues
back to top
Kit Contents
GenElute mRNA From Total RNA Kits
  • Water, Molecular Biology Grade
  • 2x Binding Solution
  • Oligo (dT) Polystyrene Beads
  • Wash Solution
  • Elution Solution
  • Spin Columns with Tubes
  • Collection Tubes
GenElute Direct mRNA Kits
  • Lysis Solution
  • Proteinase K and Hydration Reagent
  • 5 M NaCl
  • Oligo (dT) Polystyrene Beads
  • Wash Solutions 1 & 2
  • Elution Solution
  • Filtration Columns with Tubes
  • Spin Columns with Tubes
  • Collection Tubes
back to top
Preparation Time Comparison
GenElute mRNA Prep Time
Figure 1. Comparison of the preparation time, to isolate mRNA from total RNA or direct from cells or tissues, using GenElute mRNA Miniprep Kit and other commercially available kits. Each kit was prepared according to the procedure supplied by the vendor.
back to top
RNA Yield
 Human mRNA Isolation from Total RNA
Figure 2. Total RNA was prepared from Hek293 cells by a silica-binding method, using TriReagent®. mRNA was subsequently prepared from total RNA preparations using commercially available kits, following the procedures supplied by each vendor. RNA yield was measured using the RiboGreen™ RNA Quantitation Kit (Molecular Probes).
back to top
Specific mRNA Yield
GenElute Specific mRNA Yield
Figure 3. mRNA was prepared from total RNA as in Figure 2, and twenty percent of each preparation was analyzed by Northern blot hybridization with 32P-labeled probes. Hybridization to human HGPRT, human p53, and mouse p53 mRNAs were quantified with a Perkin Elmer Instant Imager.
back to top
mRNA—Suitable for Northern Blots
GenElute mRNA Northern Blot
Figure 4. Northern blot comparison of mRNA prepared with GenElute mRNA kits from other suppliers. Duplicate mRNA samples were prepared from 5 x 106 Hek293 cells (top left panels) or 25-35 mg mouse liver (top right panels) with Sigma's GenElute Direct mRNA Miniprep Kit (S) or with another commercially available direct mRNA miniprep kit (A, D, I, P, & Q). An optional release and rebind procedure (described in the Manual was included for preparation S+: RNA was hybrid-captured onto oligo dT beads, released into fresh lysis solution, and re-captured onto the same beads before washing and eluting mRNA. The release and rebinding steps were omitted for S- & Sx2 preparations. Sx2 preparations were repurified after the final elution with fresh oligo dT beads (as described in the Manual. Equal portions of each mRNA preparation, equal to the amount from 1 x 106 cells or 10 mg liver, were evaluated by Northern blot hybridization with 32P-labeled RNA probes for p53, hypoxanthine-guanine phosphoribosyl transferase (HGPRT), c-myc, and/or glyceraldehyde 3-phosphate dehydrogenase (GAPDH) mRNA.
For the bottom panels, total RNA was first prepared from Hek293 cells and from mouse liver. Duplicate mRNA samples were then prepared from 100 µg of these total RNA preparations with Sigma's GenElute mRNA Miniprep Kit (S) or with another commercially available mRNA miniprep kit (Q, A, I, P, D). Samples Sx2 were repurified after elution. Twenty percent of each preparation was evaluated by Northern blot as above. In lanes T, 5 and 2 µg of the original total RNA from cells or 10 and 5 µg of total RNA from liver were analyzed for comparison.
back to top
Amplification of mRNA from GenElute mRNA kit for cotton   Amplification of mRNA from GenElute mRNA kit for cotton
RT PCR data shown. Lane (1) Wide range DNA marker D-7058; (2) No Template Control; (3) GenElute mRNA – with no DNAse Treatment; (4) GenElute mRNA – with DNAse Treatment.
Amplification of mRNA from GenElute mRNA kit for tobacco   Amplification of mRNA from GenElute mRNA kit for tobacco
RT PCR data shown. Lane (1) Wide range DNA marker D-7058; (2) No Template Control; (3) GenElute mRNA – with no DNAse Treatment; (4) GenElute mRNA – with DNAse Treatment.
GenElute™ kit was used to extract the mRNA from chilli leaves   Genelute Kit (Product No. MRN10) was used to extract the mRNA from chilli leaves
Lane 1 & 7 has wide range marker D7058, Lane 2 & 3 mRNA preparation was shown to have no DNA contamination with no DNAse treatment as well as with Dnase treatment, lane 3 & 4 are PCR positive controls using 1 µg and 100 ng of leaf DNA as templet, Lanes 9 and 10 are showing results of RbcL geneRT PCR amplification mRNA without any DNAse or with DNAse treatment. The size of DNA and RNA amplicon is 386 bp.
GenElute™ kit was used to extract the mRNA from maize leaves   Genelute Kit (Product No. MRN10) was used to extract the mRNA from maize leaves
Lane 1 & 7 has wide range marker D7058, Lane 2 & 3 mRNA preparation was shown to have no DNA contamination with no DNAse treatment as well as with Dnase treatment, lane 3 & 4 are PCR positive controls using 1 µg and 100 ng of leaf DNA as tempelets, Lanes 9 and 10 are showing RbcS gene RT PCR amplification of mRNA with out any DNAse or with DNAse treatment. The size of DNA amplicon is 601 bp and RNA amplicon is 438 bp.
GenElute™ kit was used to extract the mRNA from sugarcane leaves   Genelute Kit (Product No. MRN10) was used to extract the mRNA from sugarcane leaves
Lane 1 & 7 has wide range marker D7058, Lane 2 & 3 mRNA preparation was shown to have no DNA contamination with no DNAse treatment as well as with Dnase treatment, lane 3 & 4 are PCR positive controls using 1 µg and 100 ng of leaf DNA as tempelets, Lanes 9 and 10 are showing Actin gene RT PCR amplification of mRNA with out any DNAse or with DNAse treatment. The size of DNA amplicon is ~2030 bp and RNA amplicon is 1057 bp.
back to top