Sigma-Aldrich
Life Science > Molecular Biology > DNA & RNA Purification > mRNA
DNA & RNA Purification
mRNA
Click on the product for more information
Product Yield Speed Starting Material Product Description Technology Applications
GenElute™ Direct mRNA Miniprep Kit Up to 5 µg 60 minutes Up to 107 mammalian cells or
50 mg of tissue per prep
Isolation of mRNA from mammalian cells and tissues Hybrid-capture onto oligo dT-polystyrene beads; wash & elute in a spin column format RT-PCR, northern blots and microarray analysis
GenElute™ mRNA Miniprep Kit 1-5% of starting total RNA 40 minutes 5-500 µg total RNA Isolation of mRNA from total RNA Hybrid-capture onto oligo dT-polystyrene beads; wash & elute in a spin column format RT-PCR, northern blots and microarray analysis