Purified PCR products are suitable for Automated Sequencing
Sequence was resolved on an ABI 310 from a purified, 645 bp corn leaf PCR product. The PCR product was purified with the GenElute PCR Clean-Up Kit. The sequence was obtained by using ABI BigDye™ Terminator Chemistry and the same primers as for the original PCR.
Close Window