Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Molecular Biology > DNA & RNA Purification > SigmaSpin™ Post-Reaction Clean-Up Plates and Columns
DNA & RNA Purification
SigmaSpin™ Post-Reaction Clean-Up Plates and Columns
SigmaSpin Post-Reaction Clean-Up Columns
SigmaSpin Sequencing Reaction Clean-Up Columns are ideal for lower throughput applications, such as clean up of probe labeling reactions or small numbers of sequencing reactions.
These columns can accept sample volumes up to 100 µl. Each column comes with a collection tube to collect the DNA during centrifugation.
SigmaSpin 96-Well Sequencing Reaction Clean-Up Plates
SigmaSpin 96-Well Sequencing Reaction Clean-Up plates provide a fast, simple, and highly efficient method for removing unincorporated dyes, excess salts, and other interfering reaction components. Each plate is packed with a size-exclusion resin, prehydrated with molecular biology grade water. Unique plate design incorporates drip directors to eliminate contamination between samples. SigmaSpin has been tested in high-throughput genome centers and core facilities with ABI Prism 3700, 3100, 377 and Amersham Biosciences MegaBACE automated DNA sequencers. SigmaSpin is compatible with all standard terminator chemistries including BigDye, DYEnamic ET and Thermo Sequenase II.
SigmaSpin Sequencing Reaction Clean-Up
SigmaSpin Plates and Columns
SigmaSpin 96-Well Clean-Up Plate
Order Information
Product Code Product Name Size
S5059 SigmaSpin Post-Reaction Clean-Up Columns 70 each
S4309 SigmaSpin 96-Well Sequencing Reaction Clean-Up Plates 2 each
back to top
Beneficial Features
Versatile
Compatible with all standard terminator chemistries and all commercially available DNA sequencers
Fast
Quick and easy protocol requires 6 minutes total centrifugation
High Performance
Pre-qualified size-exclusion resin guarantees optimum performance
Recovery
Load and recover sample volumes down to 5 µl
Precision
Unique drip directors prevent cross-contamination between samples
Return to Top
Sample Data
SigmaSpin Removes Interfering Reaction Components and Eliminates Dye Blobs
Figure 1. Sequencing reaction was purified using a SigmaSpin 96-well plate. pFLAG™ MAC plasmid (E 5644) was sequenced using BigDye™ Terminator v 3.0 chemistry (20 µl total reaction volume; BigDye premix diluted 1:1 with SeqSaver™ (S3938)). Data was generated on an ABI Prism. 3700 DNA Analyzer with POP-6™ polymer and 10X CE Buffer (B4930). Phred 20 > 600.
Click Image to View Sequence in its Entirety
Return to Top