Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
DNA & RNA Purification
RNA Reagents
Click on the product for more information
Product Yield Speed Starting Material Product Description Technology Applications
Amplification Grade DNase I N/A N/A N/A Eliminate DNA from RNA preparations N/A cDNA synthesis and RT-PCR
RNAlater™ Storage Solution N/A N/A N/A Stablilizes and protects cellular RNA for up to a month at 4°C or indefinitely at -20°C N/A mRNA and total RNA isolation, histology and immunocytochemistry