Life Science > Molecular Biology > DNA & RNA Purification > Sequencing Clean-Up
DNA & RNA Purification
Sequencing Clean-Up
Click on the product for more information
Product Speed Starting Material Product Description Technology Applications
UltraClear™ Sequencing Reaction Clean-Up Kit Flexible based on user interface ≤10 µl sequencing reaction Removes unincorporated dye terminators, including BigDye version 3.1, dNTPs, residual salts and other interfering components from sequencing reactions Ultrafiltration membrane purification in a centrifugation format Fluorescent Sequencing
SigmaSpin™
96-Well Post Reaction Clean-Up Plates
< 8 minutes 10 to 20 µl sequencing reaction Removes unincorporated dyes, including Big Dye 3, excess salts and other interfering components from sequencing reactions Resin based purification in a centrifugation format Fluorescent Sequencing
SigmaSpin™
Post Reaction Clean Up Columns
< 8 minutes 10 to 20 µl sequencing reaction Removes unincorporated dyes, including BigDye 3, excess salts and other interfering components from sequencing reactions Resin based purification in a centrifugation format Fluorescent Sequencing and Microarray Analysis