| Product |
Speed |
Starting Material |
Product Description |
Technology |
Applications |
| UltraClear™ Sequencing Reaction Clean-Up Kit |
Flexible based on user interface |
≤10 µl sequencing reaction |
Removes unincorporated dye terminators, including BigDye version 3.1, dNTPs, residual salts and other interfering components from sequencing reactions |
Ultrafiltration membrane purification in a centrifugation format |
Fluorescent Sequencing |
SigmaSpin™ 96-Well Post Reaction Clean-Up Plates |
< 8 minutes |
10 to 20 µl sequencing reaction |
Removes unincorporated dyes, including Big Dye 3, excess salts and other interfering components from sequencing reactions |
Resin based purification in a centrifugation format |
Fluorescent Sequencing |
SigmaSpin™ Post Reaction Clean Up Columns |
< 8 minutes |
10 to 20 µl sequencing reaction |
Removes unincorporated dyes, including BigDye 3, excess salts and other interfering components from sequencing reactions |
Resin based purification in a centrifugation format |
Fluorescent Sequencing and Microarray Analysis |