| GenElute™ Mammalian Total RNA Miniprep Kit |
Up to 150 µg of total RNA |
30 minutes |
Up to 107 mammalian cells or 40 mg of tissue per prep |
Isolation of total RNA from mammalian cells and tissues |
Silica bind & elute in a spin column format |
RT-PCR, northern blots and microarray analysis |
| TRI® Reagent RNA Isolation Reagent |
5 - 15 µg per million cells, 1 - 10 µg/mg tissue |
90 minutes |
Up to 100 mg tissue, 107 cells, 102 cm plate area or 0.25 ml fluid and blood derivatives |
Isolation of total RNA from a variety of starting materials |
Acid guanidine thiocyanate phenolchloroform extraction |
RT-PCR, northern blots and microarray analysis |