|
DNA & RNA Purification
UltraClear™ Sequencing Reaction Clean-Up Kit
|
The Advantage is Clear : Sigma’s UltraClear™ Sequencing Reaction Clean-Up Kit
|
The UltraClear Sequencing Reaction Clean-Up Kit offers a rapid and simple method for the clean-up of DNA sequencing reactions in a 96-well format. The UltraClear plate uses ultrafiltration membranes to separate low molecular weight contaminants,such as unincorporated dye terminators, dNTPs, and residual salts from the sequencing reaction products.
|
|
UltraClear Protocol
|
|
Dilute Sequencing Reactions
Load Ultra Clear Plate
Centrifuge
Add 40 µl Sequencing Solution
Resuspend the Purified Sequencing Products
Transfer Sequencing Product to an Appropriate Injection Plate for each configuration.
|
|
|
Features and Benefits
- Excellent quality – PHRED q>20 scores greater than 850 bases (See Data)
- Scientifically engineered to use less BigDye® version 3.1 chemistry and save money
- Optimized centrifugation procedures for maximum flexibility
- Partial 96-well plate can be used and stored at room temperature
|
|
|
Order Information
| Product Code |
Product Name |
Size |
| UC9601 |
UltraClear Sequencing Reaction Clean-Up |
1 x 96 |
| UC9604 |
UltraClear Sequencing Reaction Clean-Up |
4 x 96 |
back to top
High Quality Sequence Data with UltraClear Kit
back to top
Excellent PHRED q>20 Scores Compared to Competition
|
|
Data quality analysis based on PHRED q>20 scores. Samples analyzed on the ABI Prism® 3700 under standard run conditions. M13 template was prepared using 1/8 BigDye Terminator v3.1.
|
back to top
Optimized Protocol Accommodates a Variety of Spin Forces and Times
back to top back to DNA and RNA Purification