Sigma-Aldrich
Sigma - Life Science Logo
Life Science > Molecular Biology > Epigenetics
Molecular Biology
Epigenetics
Sigma has a wide portfolio of products to support Epigenetics research. The Imprint® Bisulfite Modification Kit is a rapid method to determine the methylation status of CpG islands. Sigma also has products to study Histone modification, including transcription regulators such as anti-histone antibodies.
Products:
Tool Box:
Applications Center:
Learning Center:
Information & News
Simplify! Global DNA Methylation Simplified.
 Imprint Methylated DNA Quantification
technology
A rapid and reliable method to measure global DNA methylation shifts in about half of a day.
  • Fast - The total protocol can be completed in about 4 hours
  • Flexible - 8 well assay strips may be used for high-throughput applications or manual analysis; genomic DNA from cultured cells, tissues, plasma, or other body fluid samples may be used
  • Convenient - This simple protocol uses a familiar ELISA-like procedure:  colorimetric quantification and no radioactivity; low DNA input from 10-200 ng
  • Complete - Includes methylated control DNA and all necessary reagents except water
  • Sensitive - Detection limit is 5 ng of fully methylated genomic DNA (we used human in our tests).
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Prestige Antibodies
 Life Science Innovations Conservation International Banner
LSI: Volume 7 Issue 1
  • Histone/Protein Acetylation: Gene Regulation and Cancer Therapy (371 K PDF)
Cancer Origins and Carcinogenesis
  • Imprint® Chromatin Immunoprecipitation Kit
  • Imprint DNA Modification Kit