DNA yields of 25-30 µg can be isolated from up to 2x106 cells or 25mg of tissue in less than 40 minutes. All reagents, columns and tubes provided.
G1N10 10 Purifications G1N70 70 Purifications G1N350 350 Purifications
For additional information visit the GenElute Mammalian DNA Miniprep Kit page |
|
DNA may be isolated from fresh or aged blood in less than 40 minutes. All contaminants are removed by provided prewash solution, an Rnase solution is provided as well.
NA2000 10 Purifications NA2010 70 Purifications NA2020 350 Purifications
For additional information visit the GenElute Blood Genomic DNA Kit page |