|
| The Imprint® Methylated DNA Quantification technology provides a rapid and reliable method to measure global DNA methylation shifts. Compared to the conventional methods of analyzing hypo- and hyper- global methylation like reverse HPLC, MALDI-TOF-MS, TLC, etc. Sigma’s Imprint®. Methylated DNA Quantification Kit (MDQ1), allows researchers to spend less time in the lab, with less hazardous components and still get highly accurate results. |
|
Benefits of the kit
ELISA based format
- Procedure complete in 4 easy to follow steps (Fig 1)
- All reagents including control methylated DNA included in the kit.
Quickly measures changes in DNA methylation
- Detection limit is 5ng of fully methylated DNA
- Input DNA can be as low as 10ng-200ng (Fig 2, Fig 3)
Complete, flexible and fast
- Process complete in 3.5 hours
- 96 well format allows option of studying single sample or high-throughput studies
Extended range of DNA sample source
- Can be used with cells, tissues, plasma and other body fluids
|
MDQ1 Standard Cruve, Methylated Control |
| ng DNA |
A450 |
Error |
| 200 |
0.594 |
0.047 |
| 100 |
0.39 |
0.03 |
| 50 |
0.251 |
0.018 |
| 25 |
0.144 |
0.009 |
| 12.5 |
0.072 |
0.0112 |
| 0 |
0 |
0 |
Calculations
To calculate relative methylation levels compared to the control DNA or DNA from a normal sample, use the following steps.
Direct Calculation Percent methylation compared to the control DNA
- Average all control, sample, and blank absorbance replicates.
- Subtract the average blank value from the average control and sample values.
- Divide the background subtracted average sample by the background subtracted average normal sample.
- Multiply by 100.
- Sample calculation:
Average Blank Absorbance = (A450) = 0.090 Average Absorbance for 60 ng control DNA = 0.785 Average Absorbance for 60 ng sample DNA = 0.654
0.654 - 0.090 x 100 = 81.15% 0.785 - 0.090
The sample DNA has a global methylation level that is 81.15% of the control DNA.
Calculation from a standard curve Percent methylation relative to DNA used to generate the standard curve
- Determine the concentration of DNA in the sample(s) by A260 or fluorimetric means.
- Create a standard curve by plotting background subtracted A450 versus control DNA (see example)
- For each background subtracted sample reading, calculate the % methylation as below:
Methyl DNA (ng) = [A450 background subtracted – intercept] / slope ng in sample
Note: in many or most cases the intercept will be zero.
View the Imprint® Methylated DNA Quantification Troubleshooting Questions
View the poster presented at ASHG, Nov 12-1, 2008 at Philadelphia "Method to Measure Global DNA Methylation"
Azacytadine, a known demethylating agent, was added directly to culture media. Cells were harvested after 48 hours. DNA was isolated using GenElute and global methylation was measured. 40 ng of DNA from treated and non-treated CHO cells was analyzed.
|
Methylation status of two lots of Sss I treated DNA was measured by LC-MS. Varying amounts of DNA was also analyzed via the MDQ1 kit. As demonstrated by the graph, the difference in methylation states can easily be detected and the signal generated is proportional to the amount of DNA used in the procedure.
|
Ordering information
| Prod No. |
Product Name |
| MDQ1- 48 Rxn |
Imprint® Methylated DNA Quantification Kit |
| MDQ1- 96 Rxn |
Imprint® Methylated DNA Quantification Kit |
Products to Support the Epigenetics Workflow using this kit
DNA extraction and modification
| Prod No. |
Product Name |
| G1N10 |
GenElute™ Mammalian Genomic DNA Miniprep Kit |
| NA2000 |
GenElute™ Blood Genomic DNA Kit |
| G2N10 |
GenElute™ Plant Genomic DNA Miniprep Kit |
| MOD50 |
Imprint® DNA Modification Kit |
DNA-protein interaction & ChIP validated antibodies
| Prod No. |
Product Name |
| CHP1 |
Imprint® Chromatin Immunoprecipitation Kit |
| A5463 |
Anti-Acetylated Proteins antibody produced in rabbit |
| H0913 |
Monoclonal Anti-acetyl-Histone H3 (Ac-Lys9) antibody produced in mouse |
| H2663 |
Monoclonal Anti-Histone Deacetylase 2 (HDAC2) antibody produced in mouse |
| H7162 |
Anti-methyl-Histone H3 (Me-Lys9) antibody produced in rabbit |
| H7789 |
Anti-acetyl-Histone H1.4 (Ac-Lys26) antibody produced in rabbit |
| H9161 |
Anti-acetyl- & phospho-Histone H3 (Ac-Lys9, pSer10) antibody produced in rabbit |
| H9286 |
Anti-acetyl-Histone H3 (Ac-Lys9) antibody produced in rabbit |
| R4279 |
Anti-RbAp46, C-terminal antibody produced in rabbit |
Mission shRNA for functional genomic studies
| Prod No. |
Product Name |
| SH3011 |
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock Epigenetic Regulators |
| SH3021 |
MISSION® shRNA Human Gene Family Set, DNA Epigenetic Regulators |
| SH3031 |
MISSION® shRNA Human Gene Family Set, Lentiviral Particles Epigenetic Regulators |
|