Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Life Science > Molecular Biology > Learning Center > Protocols > Immunoprecipitated Chromatin
Molecular Biology
Immunoprecipitated Chromatin
cshlp-small Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols.
View the protocol on BioSupplyNet.com.
Selected Protocol from "Proteins and Proteomics: A Laboratory Manual,", David R. Goodlett, Eugene C. Yi, and Philippe Mottay.
View the protocol "PCR-Based Analysis of Immunoprecipitated Chromatin" at BioSupplynet.com
Alexandre Wagschal, Katia Delaval, Maëlle Pannetier, Philippe Arnaud, and Robert Feil - Institute of Molecular Genetics, CNRS and University of Montpellier, 34293 Montpellier, France
ABSTRACT
After chromatin immunoprecipitation (ChIP), different PCR-based approaches can be used to determine how much DNA is precipitated at a locus of interest. Real-time PCR amplification is often the preferred technique. One can also use duplex PCR amplification, which is the coamplification of a fragment from the region of interest and a control fragment (e.g., the actin gene, or the tubulin gene). This approach allows for estimating relative levels of specific histone modifications along chromosomal domains. For allele-specific studies (for instance, on dosage-compensation mechanisms or on genomic imprinting), electrophoretic detection of single-strand conformation polymorphisms (SSCP) or similar strategies such as hot-stop PCR can differentiate PCR products that represent the silent allele from those amplified from the active allele. If a polymorphic restriction site is present in one allele and absent in the other, the method of choice is hot-stop PCR. If no polymorphic restriction sites are available, but there are single nucleotide polymorphisms (SNPs) that distinguish the alleles of interest, the best approach is to separate the PCR products derived from the two different alleles using SSCP. In SSCP, it is possible to discriminate denatured PCR products derived from one allele or the other because the secondary structure of each single strand will be directly dependent on the sequence itself. Hence, in nondenaturing gel conditions, each single strand will migrate differently. These four PCR-based methodologies to analyze immunoprecipitated chromatin (real-time PCR, duplex PCR, hot-stop PCR, and SSCP) are presented here.
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
100-bp DNA step ladder D5042 123 bp Ladder, buffered aqueous solution
Acrylamide solution for SSCP gels (2X) 01697 Acrylamide solution, BioChemika, for molecular biology, 40% in H2O
Acrylamide/bisacrylamide (29:1 ratio; 40% stock solution) A2792 Acrylamide/Bis-acrylamide, 29:1 (ratio), DNase, RNase and protease, none detected, Molecular Biology
Agarose A9539 Agarose, for routine use
Ammonium persulfate (APS; 10%, w/v) A9164 Ammonium persulfate, for molecular biology, powder, ≥98%
dNTPs DNTP100 Deoxynucleotide Set, 100 mM
Xylene cyanol FF X4126 Xylene Cyanol FF for molecular biology
Bromophenol blue B5525 Bromophenol Blue sodium salt for electrophoresis
Ethidium bromide solution E1385 Ethidium bromide solution, for molecular biology, 500 µg/mL in H2O, suitable for gel electrophoresis and DNA isolation procedures
PCR amplification buffer (with Taq) View the entire line of PCR amplification mixes
Restriction endonuclease View the entire line of restriction endonucleases
Restriction endonuclease buffer (10X) (supplied with the restriction endonuclease) View the entire line of restriction endonuclease buffer
Formamide F7508 Formamide, for molecular biology, ≥99.5% (GC), liquid
NaOH 72068 Sodium hydroxide solution, BioChemika Ultra, for molecular biology, 10 M in H2O
Taq DNA polymerase (5 U/µl) D1806 Taq DNA Polymerase from Thermus aquaticus, with 10× reaction buffer containing MgCl2, recombinant, expressed in Escherichia coli
TBE buffer T4073 Tris-Borate-EDTA buffer, working solution, Biotechnology Performance Certified
Template DNA D4642 Deoxyribonucleic acid from human placenta, Genomic
back to top
View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.