Molecular Biology
Competent Cell Preparation
|
The following Cold Spring Harbor Laboratory Press protocols are brought to you by BioSupplyNet in a partnership with Sigma-Aldrich. A list of products is available to use in each protocol.
View the protocol on BioSupplyNet.com.
|
ABSTRACT At its best, this method for preparing competent E. coli from Inoue et al. (1990) can challenge the efficiencies achieved by Hanahan (1983). However, under standard laboratory conditions, efficiencies of 1 x 108 to 3 x 108 transformed colonies/mg of plasmid DNA are more typical. The advantages of the procedure are that it is less finicky, more reproducible, and therefore more predictable than the original Hanahan method. This protocol differs from other procedures in that the bacterial culture is grown at 18°C rather than the conventional 37°C. Otherwise, the protocol is unremarkable and follows a fairly standard course. Why growing the cells at low temperature should affect the efficiency of transformation is anybody's guess. Perhaps the composition or the physical characteristics of bacterial membranes synthesized at 18°C are more favorable for uptake of DNA, or perhaps the phases of the growth cycle that favor efficient transformation are extended. Incubating bacterial cultures at 18°C is a challenge. Most laboratories do not have a shaking incubator that can accurately maintain a temperature of 18°C summer and winter. One solution is to place an incubator in a 4°C cold room and use the temperature control to heat the incubator to 18°C. Alternatively, there is almost no loss of efficiency if the cultures are grown at 20–23°C, which is the ambient temperature in many laboratories. Cultures incubated at these temperatures grow slowly with a doubling time of 2.5 to 4 hours. This can lead to frustration, especially late at night when it seems that the culture will never reach the desired OD600 of 0.6. The answer to this problem is to set up cultures in the evening and harvest the bacteria early the following morning. The procedure works well with many strains of E. coli in common use in molecular cloning, including XL1-Blue, DH1, JM103, JM108/9, DH5a, and HB101.
Products Available for this Protocol
| Protocol Material Description |
Product # |
Product Name |
Add to Cart |
| Buffers and Solutions |
|
|
|
| DMSO |
D8418 |
Dimethyl sulfoxide |
|
| 55 mM MnCl2.4H2O |
M8054 |
Manganese chloride tetrahydrate |
|
| 15 mM CaCl2.2H2O |
C8106 |
Calcium chloride |
|
| 250 mM KCl |
P9541 |
Potassium chloride |
|
| 10 mM PIPES (0.5M, pH 6.7) |
P1851 |
PIPES |
|
| Media |
|
|
|
| LB (Luria-Bertani) Medium |
L2542 |
LB Broth |
|
| Tryptone |
T9410 |
Tryptone, Pancreatic digest of casein |
|
| Yeast Extract |
Y1625 |
Yeast extract |
|
| NaCl |
S3014 |
Sodium chloride, for molecular biology |
|
| KOH |
P5958 |
Potassium hydroxide |
|
| H2O |
W4502 |
Water |
|
| SOB Medium |
H8032 |
Hanahan's Broth |
|
| MgSO4 |
M2643 |
Magnesium sulfate |
|
| SOC Medium |
S1797 |
SOC Medium |
|
| 20 mM glucose |
G5400 |
D-(+)-Glucose |
|
| NaOH |
72068 |
Sodium hydroxide solution |
|
Competent Cells that can be used in place of this protocol
| Product Code |
Product Name |
Add to Cart |
| BL21 Competent Cells, Uni-pack |
| B2685 |
BL21 Competent Cells, Uni-pack T1R |
|
| B2935 |
BL21 Competent Cells, Uni-pack (DE3)-T1R |
|
| B3310 |
BL21 Competent Cells, Uni-pack (DE3) pLysS-T1R |
|
| B3435 |
BL21 Competent Cells, Uni-pack (DE3) pLysE-T1R |
|
| GC10™ Competent Cells |
| G2919 |
GC10™ Competent Cells Uni-pack |
|
| G2794 |
GC10™ Competent Cells Standard Aliquots |
|
| GC5™ Competent Cells |
| G3169 |
GC5™ Competent Cells Uni-pack |
|
| G3044 |
GC5™ Competent Cells Standard Aliquots |
|
| G7419 |
GC5™ Competent Cells, 96-well |
|
| Other Competent Cells |
| H3788 |
HB101 Competent Cells, Uni-pack |
|
| J3895 |
JM109 Competent Cells, Uni-pack |
|
back to top View all available Cold Spring Harbor Laboratory Press Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.