Sigma-Aldrich
Life Science > Molecular Biology > Learning Center > Protocols > Ethidium Bromide Removal
Molecular Biology
Ethidium Bromide Removal
cshlp-small Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols.
View the protocol on BioSupplyNet.com.
Selected Protocol from "Molecular Cloning: A Laboratory Manual", Third Edition, Joseph Sambrook, David W. Russell.
Removal of Ethidium Bromide from DNA by Extraction with Organic Solvents
Joseph Sambrook
Peter Maccallum Cancer Institute and The University of Melbourne, Australia
David W. Russell
University of Texas Southwestern Medical Center, Dallas
ABSTRACT
Ethidium bromide is usually removed from DNA purified through a CsCl gradient by repeated extraction with organic solvents. The CsCl is subsequently removed by dialysis or by precipitation. This method is highly effective in removing ethidium bromide from DNA purified by equilibrium centrifugation in CsCl-ethidium bromide gradients.
Molecular Cloning Cover
Buy this book, M8265
Products Available for this Protocol
Protocol Material Description Product #  Product Name Add to Cart
Buffers and Solutions      
Ethanol E7023 Ethanol
Isoamyl alcohol or n-butanol, saturated with H2O B7906 1-Butanol
Phenol P1037 Phenol
Phenol:chloroform (1:1 v/v) P1944 Phenol:Chloroform 5:1
TE (pH 8.0) T9285 Tris-EDTA Buffer 100× Concentrate
Special Equipment      
Dialysis tubing D9277 Dialysis Tubing Cellulose Membrane
Dialysis clips Z371092 Dialysis tubing closures
back to top
View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.