| Protocol Material Description |
Product # |
Product Name |
Add to Cart |
| Alkaline Lysis Solution I |
|
|
|
| 50 mM glucose |
G5400 |
D-(+)-Glucose |
|
| 25 mM Tris-Cl (pH 8.0) |
T5941 |
Trizma® hydrochloride for molecular biology, Biotechnology Performance Certified |
|
| 10 mM EDTA (pH 8.0) |
E7889 |
Ethylenediaminetetraacetic acid disodium salt solution for molecular biology |
|
| Alkaline Lysis Solution II |
|
|
|
| 0.2 N NaOH (freshly diluted from a 10 N stock) |
72068 |
Sodium hydroxide solution, BioChemika Ultra, for molecular biology |
|
| 1% (w/v) SDS |
L4390 |
Sodium dodecyl sulfate for molecular biology |
|
| Alkaline Lysis Solution III |
|
|
|
| 5 M potassium acetate, 60.0 ml |
P1190 |
Potassium acetate for molecular biology |
|
| Glacial acetic acid, 11.5 ml |
A6283 |
Acetic acid, ReagentPlus™ |
|
| H2O, 28.5 ml |
W4502 |
Water, Molecular Biology Reagent |
|
| General Reagents |
|
|
|
| Chloramphenicol (34 mg/ml) |
C1863 |
Chloramphenicol Biotechnology Performance Certified |
|
| Ethanol |
E7023 |
Ethanol, 200 proof (absolute), for molecular biology |
|
| Isopropanol |
I9516 |
2-Propanol for molecular biology, >99% |
|
| STE |
|
|
|
| 10 mM Tris-Cl (pH 8.0) |
T5941 |
Trizma® hydrochloride for molecular biology, Biotechnology Performance Certified |
|
| 0.1 M NaCl |
S3014 |
Sodium chloride |
|
| 1 mM EDTA (pH 8.0) |
E7889 |
Ethylenediaminetetraacetic acid disodium salt solution for molecular biology |
|
| TE (pH 8.0) |
|
|
|
| 100 mM Tris-Cl (desired pH) |
T5941 |
Trizma® hydrochloride for molecular biology, Biotechnology Performance Certified |
|
| 10 mM EDTA (pH 8.0) |
E7889 |
Ethylenediaminetetraacetic acid disodium salt solution for molecular biology |
|
| Enzymes |
|
|
|
| Lysozyme (10 mg/ml) |
L3790 |
Lysozyme from chicken egg white |
|
| Media |
|
|
|
| LB |
L2897 |
LB Agar, powder |
|
| YT |
95768 |
YT Agar, BioChemika, for microbiology |
|
| Terrific Broth |
T5574 |
Terrific Broth Liquid Medium |
|