Molecular Biology
Purification of PCR Products
 |
Sigma® has prepared product lists for commonly used protocols from Cold Spring Harbor Protocols. |
View the protocol on BioSupplyNet.com.
|
Selected Protocol from "Molecular Cloning: A Laboratory Manual", Third Edition, Joseph Sambrook, David W. Russell.
Purification of PCR Products in Preparation for Cloning
Joseph Sambrook Peter Maccallum Cancer Institute and The University of Melbourne, Australia David W. Russell University of Texas Southwestern Medical Center, Dallas
ABSTRACT The residual enzymatic activity of thermostable DNA polymerases that survive the rigors of PCR can compromise subsequent enzymatic reactions. This protocol describes how to use proteinase K to destroy thermostable enzymes and to purify amplified DNA in preparation for cloning.
|
Buy this book, M8265
|
Products Available for this Protocol
| Protocol Material Description |
Product # |
Product Name |
Add to Cart |
| Ammonium acetate (10M) |
A1542 |
Ammonium acetate |
|
| Chloroform |
C2432 |
Chloroform |
|
| Ethanol |
E7023 |
Ethanol |
|
| Phenol:chlorofrom (1:1, v/v) |
P2069 |
Phenol:Chloroform:Isoamyl Alcohol 25:24:1 Saturated with 10 mM Tris, pH 8.0, 1 mM EDTA |
|
| TE (pH 8.0) |
T9285 |
Tris-EDTA Buffer 100× Concentrate |
|
| Proteinase K |
P2308 |
Proteinase K from Tritirachium album |
|
Note 1: For Step 6 of the protocol, you may use the GenElute™ PCR Clean-Up Kit, NA1020.
Note 2: As an alternative, the entire protocol could be accomplished with procuct NA1020, GenElute™ PCR Clean-Up Kit.
back to top View all available product lists for Cold Spring Harbor Laboratory Protocols
Product Association Disclaimer: The Sigma-Aldrich products listed for this specific protocol were selected either to match or to supplement the products listed within the actual protocol. The products/reagents from Sigma-Aldrich have been qualified for usage, but may not have been validated for this specific application. Please refer to the detailed product description on the usage of specific products of interest.