Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Nucleic Acid Electrophoresis
Equipment & Accessories
Mini Horizontal Electrophoresis Units
Designed for rapid, routine electrophoresis for nucleic acids and supplied with two removable UV-transparent gel casting trays with aluminum gates for tape-free gel pouring.
Product Name Product No.
Complete Set - Includes base unit, safety lid, two casting trays with aluminum gates (one for 6 x 7 and one for 6 x 10 cm gels), and two combs (1.0 mm x 8 sample wells and 1.0 mm x 12 sample wells). Z338796
Gel Casting Tray, standard form, Size: 6 cm x 7 cm Z338990
Gel Casting Tray with Aluminum casting gates, Size: 6 cm 10 cm Z339024
Colored Loading Strips – For mini horizontal gel electrophoresis unit. Provide a colored background behind sample loading area making sample wells more visible. Z339040
Comb, Size: 1.0 mm x 8 wells Z338877
Comb, Size: 1.0 mm x 16 wells Z338893
Comb, Size: 1.5 mm x 4 wells Z338907
Comb, Size: 1.5 mm x 8 wells Z338915
Comb, Size: 1.5 mm x 12 wells Z338923
Comb, Size: 2.0 mm x 4 wells Z338958
Comb, Size: 2.0 mm x 8 wells Z338966
Comb, Size: 2.0 mm x 12 wells Z338974
Comb, Size: 2.0 mm x 16 wells Z338982
Midi Horizontal Electrophoresis Unit Set
Designed for both analytical and preparative separations of nucleic acids. These 13 cm wide units are similar to the Mini units, but with the addition of buffer circulation ports. Four comb positions allow up to 112 samples to be screened on one gel.
Product Name Product No.
Complete set – Includes base unit, safety lid, one standard casting tray for 13 x 15 cm gels, and two combs (1.0 mm x 12 sample wells and 1.0 mm x 16 sample wells). Z338818
Gel Casting Tray, standard form, Size: 13 cm x 10 cm Z339253
Gel Casting Tray with Aluminum casting gates, Size 13 cm x 10 cm Z339261
Colored loading strips, for 13 cm gel trays – For midi horizontal gel electrophoresis unit. Provide a colored background behind sample loading area making sample wells more visible. Z339296
Comb, Size: 1.0 mm x 12 wells Z339075
Comb, Size: 1.0 mm x 16 wells Z339083
Comb, Size: 1.0 mm x 20 wells Z339091
Comb, Size: 1.0 mm x 24 wells Z339105
Comb, Size: 1.5 mm x 28 wells Z339113
Comb, Size: 1.5 mm x 12 wells Z339148
Comb, Size: 1.5 mm x 16 wells Z339156
Comb, Size: 1.5 mm x 20 wells Z339164
Comb, Size: 1.5 mm x 24 wells Z339172
Comb, Size: 2.0 mm x 28 wells Z339180
Comb, Size: 2.0 mm x 10 wells Z339199
Comb, Size: 2.0 mm x 16 wells Z339210
Comb, Size: 2.0 mm x 20 wells Z339229
Comb, Size: 2.0 mm x 24 wells Z339237
Comb, Size: 2.0 mm x 28 wells Z339245
Maxi Horizontal Electrophoresis Unit Set
Designed for both analytical and preparative separations of nucleic acids with buffer circulation ports to prevent buffer depletion. Four comb positions permit the screening of up to 160 samples.
Product Name Product No.
Complete Cooled Unit- Includes base unit with hose connectors for circulating coolant, safety lid, one standard casting tray for 20 x 20 cm gels, and two combs, 1.0 mm x 16 sample wells. Z338834
Gel Casting Tray, standard form, Size: 20 cm x 20 cm Z339458
Colored gel loading strips – Provide a colored background behind the sample loading area making sample wells more visible. Z339466
Comb Size: 1.0 mm x 16 wells Z339326
Comb Size: 1.0 mm x 20 wells Z339334
Comb Size: 1.0 mm x 28 wells Z339342
Comb Size: 1.0 mm x 40 wells Z339350
Comb Size: 1.0 mm x 16 wells Z339369
Comb Size: 1.0 mm x 28 wells Z339385
Comb Size: 1.0 mm x 40 wells Z339393
Comb Size: 1.0 mm x 16 wells Z339407
Comb Size: 1.0 mm x 20 wells Z339415
Comb Size: 1.0 mm x 28 wells Z339423
Comb Size: 1.0 mm x 40 wells Z339431
Maxi-Plus Horizontal Electrophoresis Unit Set
This electrophoresis unit has been specifically designed to allow quick and convenient screening of PCR products. When using a 96 well PCR machine, all samples can be screened on a single gel. Loading is simple using a standard multichannel pipette. The unit is constructed of 12 mm thick acrylic to prevent warping and is supplied with a UV transparent tray for easy visualization. For varying sample numbers a range of alternative combs can be used. As there are 12 comb positions, up to 624 samples can be separated on a single gel.
Product Name Product No.
Complete Uncooled Unit - Includes base unit, safety lid, one standard casting tray for 25 x 30 cm gels, and two combs, 1.0 mm x 26 sample wells Z338842
Complete Cooled Unit - Includes base unit with hose connectors for circulating coolant, safety lid, one standard casting tray for 25 x 30 cm gels, and two combs, 1.0 mm x 26 sample wells Z338850
Gel Casting Tray, Standard form, Size: 25 cm x 30 cm Z339539
Comb, Size: 1.0 mm x 26 wells Z339474
Comb, Size: 1.5 mm x 26 wells Z339490
Comb, Size: 2.0 mm x 26 wells Z339512
Comb, Size: 2.0 mm x 52 wells Z339520
Transilluminators, UV
For visualization of Ethidium bromide-stained nucleic acids and similar fluorescence detection systems.
Features include:
  • Single-band and dual-band units available. All include 312 nm (midrange UV) bulbs, which corresponds with the fluorescence excitation maximum of Ethidium bromide. Fluorescence intensity is enhanced, while photobleaching and photonicking of stained nucleic acids are reduced. Dual band units also include 254 nm (short-wave UV) bulbs.
  • Six 15 W bulbs illuminate the viewing area uniformly.
  • Intensity selector: switch between 100% for reading/photographing gels and 70% for preparing/cutting gels.
  • Cooling fan: helps prevent thermal damage to both the samples and the glass filter.
  • Long life filter is highly resistant to solarization; unlimited life at 312 nm and above, 3000 hr at 254 nm.
  • Built-in hinged UV safety screen
(The 110 V units are not offered for sale or supply to EU countries. Safety equipment for UV-blocking face shields and additional UV-blocking goggles.)
Product Name Product No.
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z363677
Viewing area: 20 cm x 20 cm, Wavelength: 254-312 nm (selectable), 110V AC, 60Hz (not available in EU)
Z363782
Transilluminators, UV/White Light
One transilluminator with two functions
The left side is a 20 x 20 cm white light table for reading autoradiograms, protein gels, and multiwell plates. Two 8W color-balanced fluorescent tubes diffused through an opal glass stage uniformly illuminate it. The right side is a 20 x 20 cm 312nm UV transilluminator for Ethidium bromide-stained gels and other fluorescence applications. Illumination comes from six 8W tubes. The UV filter resists solarization indefinitely with 312 nm light source. A hinged UV safety screen protects the entire stage and can be propped up like a work shield.
Product Name Product No.
115 V AC, 60 Hz (not available in EU) Z363820