Sigma-Aldrich
Life Science > Molecular Biology > Nucleic Acid Electrophoresis > Equipment & Accessories
Nucleic Acid Electrophoresis
Equipment & Accessories
Agarose and Acrylamide | Dyes, Loading Buffers, and Nucleic Acid Gel Stains | Markers and Ladders | Running Buffers | Equipment and Accessories
Equipment and Accessories
Sigma-Aldrich offers a variety of gel boxes, power supplies and accessories for your nucleic acid electrophoresis research.
Mini Horizontal Electrophoresis Units
Midi Horizontal Electrophoresis Unit Set
Maxi Horizontal Electrophoresis Unit Set
Maxi-Plus Horizontal Electrophoresis Unit Set
Accessories
Power Supplies
Transilluminators, UV
Transilluminators, UV/White Light
Transilluminators, White Light
Mini Horizontal Electrophoresis Units
mini-horizontal-electro
Designed for rapid, routine electrophoresis for nucleic acids and supplied with two removable UV-transparent gel casting trays with aluminum gates for tape-free gel pouring.
Product Name Technical Bulletin Product #
Complete Set - Includes base unit, safety lid, two casting trays with aluminum gates (one for 6 x 7 and one for 6 x 10 cm gels), and two combs (1.0 mm x 8 sample wells and 1.0 mm x 12 sample wells). sg_dd_hts_tbicon  Z33,879-6
Gel Casting Tray, standard form, Size: 6 cm x 7 cm   Z33,899-0
Gel Casting Tray with Aluminum casting gates, Size: 6 cm x 7 cm   Z33,901-6
Gel Casting Tray with Aluminum casting gates, Size: 6 cm 10 cm   Z33,902-4
Colored Loading Strips – For mini horizontal gel electrophoresis unit. Provide a colored background behind sample loading area making sample wells more visible.   Z33,904-0
Comb, Size: 1.0 mm x 4 wells   Z33,886-9
Comb, Size: 1.0 mm x 8 wells   Z33,887-7
Comb, Size: 1.0 mm x 12 wells   Z33,888-5
Comb, Size: 1.0 mm x 16 wells   Z33,889-3
Comb, Size: 1.5 mm x 4 wells   Z33,890-7
Comb, Size: 1.5 mm x 8 wells   Z33,891-5
Comb, Size: 1.5 mm x 12 wells   Z33,892-3
Comb, Size: 1.5 mm x 16 wells   Z33,893-1
Comb, Size: 2.0 mm x 4 wells   Z33,895-8
Comb, Size: 2.0 mm x 8 wells   Z33,896-6
Comb, Size: 2.0 mm x 12 wells   Z33,897-4
Comb, Size: 2.0 mm x 16 wells   Z33,898-2
back to top
Midi Horizontal Electrophoresis Unit Set
Designed for both analytical and preparative separations of nucleic acids. These 13 cm wide units are similar to the Mini units, but with the addition of buffer circulation ports. Four comb positions allow up to 112 samples to be screened on one gel.
Product Name Technical Bulletin Product #
Complete set – Includes base unit, safety lid, one standard casting tray for 13 x 15 cm gels, and two combs (1.0 mm x 12 sample wells and 1.0 mm x 16 sample wells).  sg_dd_hts_tbicon Z33,881-8
Gel Casting Tray, standard form, Size: 13 cm x 10 cm   Z33,925-3
Gel Casting Tray with Aluminum casting gates, Size 13 cm x 10 cm   Z33,926-1
Replacement Casting gates   Z33,928-8
Colored loading strips, for 13 cm gel trays – For midi horizontal gel electrophoresis unit. Provide a colored background behind sample loading area making sample wells more visible.   Z33,929-6
Comb, Size: 1.0 mm x 10 wells   Z33,906-7
Comb, Size: 1.0 mm x 12 wells   Z33,907-5
Comb, Size: 1.0 mm x 16 wells   Z33,908-3
Comb, Size: 1.0 mm x 20 wells   Z33,909-1
Comb, Size: 1.0 mm x 24 wells   Z33,910-5
Comb, Size: 1.5 mm x 28 wells   Z33,911-3
Comb, Size: 1.5 mm x 10 wells   Z33,912-1
Comb, Size: 1.5 mm x 12 wells   Z33,914-8
Comb, Size: 1.5 mm x 16 wells   Z33,915-6
Comb, Size: 1.5 mm x 20 wells   Z33,916-4
Comb, Size: 1.5 mm x 24 wells   Z33,917-2
Comb, Size: 2.0 mm x 28 wells   Z33,918-0
Comb, Size: 2.0 mm x 10 wells   Z33,919-9
Comb, Size: 2.0 mm x 12 wells   Z33,920-2
Comb, Size: 2.0 mm x 16 wells   Z33,921-0
Comb, Size: 2.0 mm x 20 wells   Z33,922-9
Comb, Size: 2.0 mm x 24 wells   Z33,923-7
Comb, Size: 2.0 mm x 28 wells   Z33,924-5
Gel Casting Tray, Standard Form, Size: 13 cm x 15 cm   Z34,044-8
Gel Casting Tray with Aluminum casting gates   Z34,045-6
back to top
Maxi Horizontal Electrophoresis Unit Set
Midi-Horizontal-Electrophor 
Designed for both analytical and preparative separations of nucleic acids with buffer circulation ports to prevent buffer depletion. Four comb positions permit the screening of up to 160 samples.
Product Name Technical Bulletin Product #
Complete Uncooled Unit – Includes base unit, safety lid, one standard casting tray for 20 x 20 cm gels, and two combs, 1.0 mm x 16 sample wells. sg_dd_hts_tbicon
Z33,882-6
Complete Cooled Unit- Includes base unit with hose connectors for circulating coolant, safety lid, one standard casting tray for 20 x 20 cm gels, and two combs, 1.0 mm x 16 sample wells. sg_dd_hts_tbicon  Z33,883-4
Gel Casting Tray, standard form, Size: 20 cm x 20 cm   Z33,945-8
Colored gel loading strips – Provide a colored background behind the sample loading area making sample wells more visible.   Z33,946-6
Comb Size: 1.0 mm x 16 wells   Z33,932-6
Comb Size: 1.0 mm x 20 wells   Z33,933-4
Comb Size: 1.0 mm x 28 wells   Z33,934-2
Comb Size: 1.0 mm x 40 wells   Z33,935-0
Comb Size: 1.0 mm x 16 wells   Z33,936-9
Comb Size: 1.0 mm x 20 wells   Z33,934-7
Comb Size: 1.0 mm x 28 wells   Z33,938-5
Comb Size: 1.0 mm x 40 wells   Z33,939-3
Comb Size: 1.0 mm x 16 wells   Z33,940-7
Comb Size: 1.0 mm x 20 wells   Z33,941-5
Comb Size: 1.0 mm x 28 wells   Z33,942-3
Comb Size: 1.0 mm x 40 wells   Z33,943-1
back to top
Maxi-Plus Horizontal Electrophoresis Unit Set
Z338842
This electrophoresis unit has been specifically designed to allow quick and convenient screening of PCR products. When using a 96 well PCR machine, all samples can be screened on a single gel. Loading is simple using a standard multichannel pipette. The unit is constructed of 12 mm thick acrylic to prevent warping and is supplied with a UV transparent tray for easy visualization. For varying sample numbers a range of alternative combs can be used. As there are 12 comb positions, up to 624 samples can be separated on a single gel.
Product Name Technical Bulletin Product #
Complete Uncooled Unit - Includes base unit, safety lid, one standard casting tray for 25 x 30 cm gels, and two combs, 1.0 mm x 26 sample wells  sg_dd_hts_tbicon Z33,884-2
Complete Cooled Unit - Includes base unit with hose connectors for circulating coolant, safety lid, one standard casting tray for 25 x 30 cm gels, and two combs, 1.0 mm x 26 sample wells sg_dd_hts_tbicon
Z33,885-0
Gel Casting Tray, Standard form, Size: 25 cm x 30 cm   Z33,953-9
Colored gel loading strips - Provide a colored background behind sample loading area making sample wells more visible   Z33,954-7
Comb, Size: 1.0 mm x 26 wells   Z33,947-4
Comb, Size: 1.0 mm x 52 wells - Well spacing compatible with multi-channel pipettes   Z33,948-2
Comb, Size: 1.5 mm x 26 wells   Z33,949-0
Comb, Size: 1.5 mm x 52 wells   Z33,950-4
Comb, Size: 2.0 mm x 26 wells   Z33,951-2
Comb, Size: 2.0 mm x 52 wells   Z33,952-0
back to top
Accessories
Product Name Product #
Buffer Recirculation Connectors for Midi, Maxi, and Maxi-Plus Units Z33,931-8
Gel Levelling Table - With four levelling screws and a recessed bubble level (20 cm x 30 cm) Z33,955-5
Replacement Platinum Electrode Wire (50 cm x 0.2 mm) Z33,905-9
back to top
Power Supplies
Power Supply, 250 Volt, Dual Mode
Programmable Power Supply
Power Supply, 250 Volt, Dual Mode
Ideal for use with polyacrylamide or agarose gel electrophoresis and blotting:
  • Dual control modes (constant voltage or constant current) with automatic cross over
  • Integral shutdown timer (0-24 hr)
  • Reasonably priced
  • Quad output: four sets of recessed connectors
  • Integral timer: 0-24 hrs
  • Automatic shutdown of DC power when last electrophoresis unit is disconnected
  • Operating temperature: 0-30 °C
  • Size - 14 cm H x 22 cm W x 19 cm D
  • Power - 500mA+ mA, 250 V (capacity)
Product Name Product #
110V AC, US 3-pin plug (not available in EU) PS251-1
220V AC, European 2-pin plug PS251-2
Programmable Power Supply
Ideal for use with high voltage applications including IEF and DNA sequencing, but is also suitable for methods requiring less power (poly acrylamide or agarose gel electrophoresis and blotting):
  • Programmable - Can store up to 20 different 10 step programs
  • Constant voltage, constant current, and constant power modes with auto crossover
  • Capacity 300mA, 300W
  • Integral shutdown timer (0-100 hr, count up or count down)
  • Integrator capacity 999,999V-hr, 99,999mA-hr, 99,99W-hr
  • Four sets of recessed output connectors
  • Automatic shutdown on disconnect of last gel unit
  • Operating temperature range: 0-30 °C
  • Size - 14 cm H x 22 cm W x 19 cm D
  • Power - 4000 V (programmable)
Product Name Product #
110V AC, 60 Hz, US 3-pin plug (not available in EU) PS4010-1
220V AC, 50Hz PS4010-2
back to top
Transilluminators, UV
 Z363669-left-213
For visualization of Ethidium bromide-stained nucleic acids and similar fluorescence detection systems.
Features include:
  • Single-band and dual-band units available. All include 312 nm (midrange UV) bulbs, which corresponds with the fluorescence excitation maximum of Ethidium bromide. Fluorescence intensity is enhanced, while photobleaching and photonicking of stained nucleic acids are reduced. Dual band units also include 254 nm (short-wave UV) bulbs.
  • Six 15 W bulbs illuminate the viewing area uniformly.
  • Intensity selector: switch between 100% for reading/photographing gels and 70% for preparing/cutting gels.
  • Cooling fan: helps prevent thermal damage to both the samples and the glass filter.
  • Long life filter is highly resistant to solarization; unlimited life at 312 nm and above, 3000 hr at 254 nm.
  • Built-in hinged UV safety screen
(The 110 V units are not offered for sale or supply to EU countries. Safety equipment for UV-blocking face shields and additional UV-blocking goggles.)
Product Name Product #
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,366-9
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,367-7
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,368-5
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,369-3
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,378-2
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,379-0
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,380-4
Viewing area: 20 cm x 20 cm, Wavelength: 312 nm (only), 110V AC, 60Hz (not available in EU) Z36,381-2
back to top
Transilluminators, UV/White Light
Z363820-top-right-213
One transilluminator with two functions
The left side is a 20 x 20 cm white light table for reading autoradiograms, protein gels, and multiwell plates. Two 8W color-balanced fluorescent tubes diffused through an opal glass stage uniformly illuminate it. The right side is a 20 x 20 cm 312nm UV transilluminator for Ethidium bromide-stained gels and other fluorescence applications. Illumination comes from six 8W tubes. The UV filter resists solarization indefinitely with 312 nm light source. A hinged UV safety screen protects the entire stage and can be propped up like a work shield.
Product Name Product #
115 V AC, 60 Hz (not available in EU) Z36,382-0
220 V AC, 50 Hz Z36,383-9
back to top
Transilluminators, White Light
Z363707-bottom-right-213
Well suited to a wide variety of laboratory and graphics applications: viewing and photographing stained protein gels, autoradiograms, or multiwell plates; previewing slides and overhead transparencies; registering transparent overlays.
Features include:
  • 20 x 35 cm opal glass viewing area
  • Uniformity of +7% over entire area
  • Intensity selector: 70% or 100%
  • Color balanced (6500°K) white light for true color perception reduces the need for color-correcting filters
  • Cooling fan extends life of unit and prevents thermal damage to specimens.
White light transilluminators contain two 15 W and two 6 W fluorescent bulbs.
Product Name Product #
120V AC Z36,370-7
220 V AC Z36,371-5