Higher yields and more sensitivity with KlenTaq DV ReadyMixes
KlenTaq DV provides higher yields than the leading competitor's high-yield polymerase.
KlenTaq DV ReadyMixes and competitors' enzymes were used to amplify a 2.5 kb fragment of lambda DNA. Reactions contained 2.5 ng, 0.5 ng or 50 pg of lambda DNA template. Reactions assembled per suppliers' recommendations.
Lane 1: RED KlenTaq DV ReadyMix
Lane 2: KlenTaq DV ReadyMix
Lane 3: Taq DNA polymerase
Lane 4: Supplier S, enzyme Y
Lane 5: Supplier A, enzyme A
Lane M: Wide Range DNA marker
Close Window