Sigma-Aldrich
Life Science > Molecular Biology > PCR/Amplification > PCR Products
  • Pricing & availability is not currently available.
    Questions? Contact Technical Service
  • USA Home
  • Product Directory
  • Molecular Biology
  • PCR/Amplification
PCR/Amplification
Try CleanAmp dNTPs
The polymerase chain reaction (PCR) is a powerful core molecular biology technique. It is an efficient and rapid in vitro method for enzymatic amplification of specific DNA or RNA sequences from nucleic acids of various sources. A simple PCR reaction consists of a set of synthetic oligonucleotide primers that flank the target DNA sequence, target DNA, a thermostable DNA polymerase and dNTPs. A repetitive series of cycles involving template denaturation, primer annealing, followed by extension of the annealed primers, yields tremendous amounts of DNA. Because the strands synthesized in one cycle serve as a template in the next, a million-fold increase in the DNA amount is achieved in just 20 cycles.
Sigma offers a broad range of products optimized for PCR reactions. From our novel REDTaq® DNA polymerase to our PCR enhancers, each component is tested thoroughly to ensure you achieve the best results possible in your PCR reactions. Visit this site often for the most recent PCR product and technical updates.
PCR Master Mix Calculator | PCR Technical Animations
  • ReadyScript™ cDNA Synthesis Mix - (1)
  • KiCqStart™ SYBR® Green qPCR ReadyMix™ - (4)
  • CleanAmp dNTPs - (3)
  • Hot Start PCR - (9)
  • Long & Accurate PCR - (7)
  • PCR Optimization - (5)
  • PCR Reaction Components - (12)
  • Quantitative PCR - (12)
  • RT-PCR - (9)
  • Routine Amplification - (10)
  • Specialty Enzymes - (2)
  • Human Genomic DNA - (6)
  • Deoxynucleotides - (15)
  • Featured Products - (3)
  • Genomic DNA Amplification - (31)
  • PCR Books - (18)
  • PCR Lab Equipment - (411)
  • Whole Genome Amplification - (5)