Sigma-Aldrich
Life Science > Molecular Biology > PCR/Amplification > Quantitative PCR > Capillary-based qPCR
PCR/Amplification
Capillary-based qPCR
SYBR Green JumpStart Taq ReadyMix, Capillary formulation combines the advantages of a hot start enzyme, JumpStart taq, in a 2X concentrate ReadyMix specifically designed for use with capillary instruments, such as the Roche LightCycler® real–time thermal cycler.
  • Convenient – 2X concentrate ReadyMix designed specifically for use with capillary instruments.
  • Specific – JumpStart Taq antibody prevents non–specific product formation through it hot–start mechanism.
  • Optimized Formulation – Facilitate carry-over prevention from previous PCR with our specially formulated ReadyMix containing dUTP.
Product Name Technical Bulletin Product # 
SYBR Green JumpStart Taq ReadyMix for QPCR, Capillary formulation sg_dd_hts_tbicon S1816
JumpStart Taq ReadyMix with dUTP   D9191
Click Image to View Figure in its Entirety Figure 1. Efficient & Sensitive Amplification with Human Genomic DNA.
Back to Quantitative PCR home page.
Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification