Efficient & Sensitive Amplification with Human Genomic DNA
Efficient and sensitive amplification with human genomic DNA. Quantitative PCR was performed on human genomic DNA using the Roche LightCycler. The template was diluted 10–fold in subsequent wells with concentrations of 30 ng to 30 pg. SYBR Green Taq ReadyMix, capillary formulation (in red) has comparable sensitivity and efficiency to the SYBR Green master mix (in blue) from supplier R.
Close Window