KiCqStart® SYBR® Green qPCR ReadyMix™
KiCqStart your research into high gear! KiCqStart (pronounced “kickstart”) line of qPCR reagents are rigorous ReadyMixes for gene expression studies. Simple yet flexible, KiCqStart puts you in the driver’s seat.
Built for quality results…
  • Stringent hot-start mechanism for greater specificity
  • Linear up to 6 logs of input template
  • MIQE guidelines used for pre-designed primers
Linear Dynamic Range
Target gene was amplified from log-fold dilutions of total HeLa cell cDNA using KiCqStart® SYBR® Green qPCR ReadyMix™ or Competitor Q according to each manufacturer′s protocol.
…with flexible, easy-to-use mixes
  • ReadyMixes with all non-custom reaction components
  • Run experiments in conventional or fast PCR mode
  • Formats available for all major qPCR instruments
Fast or Conventional Cycling
Fast-cycling KiCqStart® SYBR® Green qPCR ReadyMix™ demonstrates equal performance to conventional SYBR Green protocol and reagents.
 Basic Process
KiCqStart ReadyMixes are ready-to-use, highly sensitive master mixes containing all required basic components for qPCR. Simply add your primers and water to complete the assay cocktail. Aliquot the cocktail across desired number of tubes or plates then add your template. KiCqStart ReadyMixes may be run in either conventional or fast PCR mode. For more details, see product usage information at KiCqStart Universal SYBR Green qPCR Protocol.
 Predesigned Primers and Gene Arrays
KiCqStart® SYBR® Green Primers are pre-designed primer pairs for gene expression. To comply with MIQE, they have been developed using sophisticated bioinformatics tools and validated in silico to avoid off-target amplification. Primer pairs were also designed to detect the most prevalent mRNA splice variant in eukaryotes for each gene, with amplicon sizes ranging between 75 and 200 base pairs. KiCqStart® SYBR® Green Primers Gene Arrays are the original KiCqStart Primers formatted into 96- or 384-well pre-plated arrays. Arrays may be customized and are available for designated research areas such as apoptosis, cell adhesion, cell cycle, cytokines and chemokines, nuclear hormone receptors, protein kinases and phosphatases.
KiCqStart Primers   KiCqStart Primers Gene Arrays
 Formatted for Your Instrument
Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagents to your specific instrument.  KiCqStart ReadyMixes contain ROX passive reference dye at concentrations recommended by these (and other) popular cyclers.
Real-time PCR Instrument Name KiCqStart® SYBR® Green qPCR ReadyMix™ KiCqStart® SYBR® Green qPCR ReadyMix™ Low Rox KiCqStart® SYBR® Green qPCR ReadyMix™ with ROX KiCqStart® SYBR® Green qPCR ReadyMix™ for iQ
  KCQS00 KCQS01 KCQS02 KCQS03
Agilent AriaMX      
Applied Biosystems 5700      
Applied Biosystems 7000      
Applied Biosystems 7300      
Applied Biosystems 7500      
Applied Biosystems 7500 Fast      
Applied Biosystems 7700      
Applied Biosystems 7900      
Applied Biosystems 7900 HT      
Applied Biosystems 7900 HT Fast      
Applied Biosystems StepOnePlus™      
Applied Biosystems StepOne™      
Applied Biosystems ViiA™ 7      
Applied Biosystems QuantStudio™      
BioRad CFX      
Bio-Rad CFX384™      
Bio-Rad® CFX96™      
BioRad iCycler iQ™      
BioRad iQ™5      
Bio-Rad MiniOpticon™      
BioRad MyiQ™      
Bio-Rad/MJ Chromo4™      
Bio-Rad/MJ Opticon 2™      
Bio-Rad/MJ Opticon™      
Cepheid SmartCycler®      
Douglas Scientific IntelliQube®      
Eppendorf Mastercycler® ep realplex      
Eppendorf Mastercycler® ep realplex2 S      
Illumina Eco™ qPCR      
Qiagen/Corbett Rotor-Gene® 3000      
Qiagen/Corbett Rotor-Gene® 6000      
Qiagen/Corbett Rotor-Gene® Q      
QIAGEN Rotor-Gene Q      
Roche LightCycler™ 480      
Stratagene Mx3000P™      
Stratagene Mx3005P™      
Stratagene Mx4000™      
 Products
Detailed KiCqStart product information can be found below. To learn more about qPCR, visit our PCR Technologies Guide, SYBR Green 1-based qPCR technical animation, or qPCR and MIQE Seminar Series.
Product No. Description
KCQS00 KiCqStart SYBR® Green qPCR ReadyMix For Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems
KCQS01 KiCqStart SYBR® Green qPCR ReadyMix Low ROX™, for ABI and Stratagene instruments
KCQS02 KiCqStart SYBR® Green qPCR ReadyMix with ROX™ for ABI instruments
KCQS03 KiCqStart SYBR® Green qPCR ReadyMix iQ™, with fluorescein for Bio-Rad systems
KCQS04 KiCqStart Probe qPCR ReadyMix for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems
KCQS05 KiCqStart Probe qPCR ReadyMix, Low ROX™ with Low ROX for ABI and Stratagene instruments
KCQS06 KiCqStart Probe qPCR ReadyMix, ROX™ with ROX for ABI instruments
KCQS07 KiCqStart One-Step Probe RT-qPCR ReadyMix for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems
KCQS08 KiCqStart One-Step Probe RT-qPCR ReadyMix, Low ROX™ with Low ROX for ABI and Stratagene instruments
KCQS09 KiCqStart One-Step Probe RT-qPCR ReadyMix, ROX™ with ROX for ABI instruments