|
|||||||||||||||||
|
|||||||||||||||||
SYBR Green I, a commonly used fluorescent DNA binding dye, binds all double–stranded DNA and detection is monitored by measuring the increase in fluorescence throughout the cycle. SYBR Green I has an excitation and emission maxima of 494 nm and 521 nm, respectively. Specificity of Sigma's SYBR based QPCR detection is greatly enhanced by the incorporation of a hot–start mediated taq polymerase, JumpStart Taq.
Mechanism of SYBR Green–based detection
Back to Quantitative PCR home page. Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification |
|||||||||||||||||
|
|
|||||||||||||||||