Sigma-Aldrich
Life Science > Molecular Biology > PCR/Amplification > Ultrapure Nucleotides
PCR/Amplification
Ultrapure Nucleotides
Sigma-Aldrich's Ultrapure Nucleotides are available as individual nucleotides or mixes. Each product is tested for suitability in PCR and DNase and RNase free.
  • Ultrapure dNTPs – Minimum ≥ 99% determined by HPLC
  • Minimize Contamination Risk – Certified DNase & RNase free
  • Convenient Pre–mixed dNTP Solution – Equimolar amounts of each dNTP means less pipetting
10mM solutions
100mM solutions
dNTP
Product
Number
Pack Size
Product
Number
Pack Size
dATP D6920 0.5 ml (5 µmol) D4788 0.25 ml (25µmol)
dCTP D7045 0.5 ml (5 µmol) D4913 0.25 ml (25µmol)
dGTP D7170 0.5ml (5 µmol) D5038 0.25ml (25µmol)
TTP T7791 0.5ml (5 µmol) T9656 0.25ml (25µmol)
dNTP Set
1 vial of
each dNTP
DNTP-10 0.5 ml (5 µmol)
of each dNTP
DNTP-100
DNTP-100A
0.25 ml (25 µmol)
of each dNTP
1.00 ml (100 µmol)
of each dNTP
dNTP Mix
Solution of all
four dNTPs
D7295 0.2 ml (2 µmol)
0.5 ml (5 µmol)
20 x 0.2ml (20x2 5mol)
PCR Home Page Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification