Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Plant Proteomics
Product Highlights
Plant Protein Extraction Kits Protein extraction kits that are specialized for the extraction and isolation of high quality protein from plant tissue samples.
CelLytic™ P: A convenient method for efficient plant cell lysis and protein solubilization.
CelLytic™ PN: For the rapid isolation of nuclei and extraction of functional nuclear proteins from plant leaves.
Chloroplast Isolation Kit: Provides a quick and efficient procedure to isolate intact chloroplasts from plant leaves.