| Component (mg/L) |
C1416 |
D6162 |
G5768 |
G5893 |
H2395 |
M6774 |
M5524 |
Q3376 |
S6765 |
W0876 |
| Aluminum chloride • 6H2O |
|
| Ammonium nitrate |
|
1416.0 |
|
|
|
400.0 |
1650.0 |
400.0 |
|
|
| Ammonium phosphate monobasic |
|
115.03 |
|
|
|
300.0 |
|
| Ammonium sulfate |
463.0 |
|
134.0 |
134.0 |
|
| Boric acid |
1.6 |
4.8 |
3.0 |
3.0 |
2.86 |
6.2 |
6.2 |
6.2 |
5.0 |
1.5 |
| Calcium chloride anhydrous |
125.33 |
112.5 |
113.24 |
113.24 |
|
72.5 |
332.2 |
|
151.0 |
|
| Calcium nitrate |
|
1367.0 |
|
|
656.4 |
386.0 |
|
833.77 |
|
200.0 |
| Calcium phosphate tribasic |
|
| Cobalt chloride • 6H2O |
|
|
0.025 |
0.025 |
|
|
0.025 |
0.025 |
0.1 |
|
| Cupric sulfate • 5H2O |
|
0.25 |
0.025 |
0.025 |
0.08 |
0.25 |
0.025 |
0.025 |
0.2 |
|
| Na2-EDTA |
37.25 |
45.4 |
37.3 |
37.25 |
|
37.3 |
37.26 |
37.3 |
20.0 |
|
| ZnNa2-EDTA |
|
| Ferric chloride • 6H2O |
|
| Ferric citrate |
|
| Ferric sulfate |
|
2.5 |
| Ferric tartrate • 2H2O |
|
5.32 |
|
| Ferrous sulfate • 7H2O |
27.85 |
33.8 |
27.8 |
27.85 |
|
27.8 |
27.8 |
27.8 |
15.0 |
|
| Magnesium sulfate |
90.37 |
361.49 |
122.09 |
122.09 |
240.76 |
180.7 |
180.7 |
175.79 |
195.4 |
360.0 |
| Manganese chloride • 4H2O |
|
1.81 |
|
| Manganese sulfate • H2O |
3.33 |
33.5 |
10.0 |
10.0 |
|
22.3 |
16.9 |
0.76 |
10.0 |
5.04 |
| Molybdenum trioxide |
|
0.016 |
|
| Molybdic acid (sodium salt) • 2H2O |
|
0.39 |
0.25 |
0.25 |
|
0.25 |
0.25 |
0.25 |
0.1 |
|
| Nickel chloride • 6H2O |
|
| Nickel sulfate • 6H2O |
|
0.005 |
|
| Potassium chloride |
|
65.0 |
| Potassium iodide |
0.8 |
|
0.75 |
0.75 |
|
|
0.83 |
0.08 |
1.0 |
0.75 |
| Potassium nitrate |
2830.0 |
|
2500.0 |
2500.0 |
606.6 |
|
1900.0 |
1800.0 |
2500.0 |
80.0 |
| Potassium phosphate monobasic |
400.0 |
265.0 |
|
|
|
170.0 |
170.0 |
270.0 |
|
|
| Potassium sulfate |
|
1559.0 |
|
|
|
990.0 |
|
| Sodium nitrate |
|
| Sodium phosphate monobasic |
|
|
130.5 |
130.5 |
|
16.5 |
| Sodium sulfate |
|
200.0 |
| Zinc nitrate • 6H2O |
|
17.0 |
|
| Zinc sulfate • 7H2O |
1.5 |
|
2.0 |
2.0 |
0.22 |
8.6 |
8.6 |
8.6 |
1.0 |
2.67 |
| |
| Organics (mg/L) |
C1416 |
D6162 |
G5768 |
G5893 |
H2395 |
M6774 |
M5524 |
Q3376 |
S6765 |
W0876 |
| Activated charcoal |
|
| Adenine hemisulfate |
|
| Agar |
|
| 6-Benzylaminopurine (BA) |
|
| D-Biotin |
|
6gamma-gamma- Dimethylallylaminopurine |
|
| Glycine (free base) |
|
| Indole-3-acetic acid |
|
| Indole-3-butyric acid |
|
| Kinetin |
|
| MES (free acid) |
|
| myo-Inositol |
|
|
|
100.0 |
|
| alpha-Naphthaleneacetic acid |
|
| Nictotinic acid (free acid) |
|
|
|
1.0 |
|
| Peptone |
|
| Pyridoxine • HCl |
|
|
|
1.0 |
|
| Sucrose |
|
| Thiamine • HCl |
|
|
|
10.0 |
|
| |
| |
C1416 |
D6162 |
G5768 |
G5893 |
H2395 |
M6774 |
M5524 |
Q3376 |
S6765 |
W0876 |
| Grams of powder to prepare 1 L |
4.0 |
5.2 |
3.1 |
3.2 |
1.6 |
2.3 |
4.3 |
3.6 |
3.2 |
0.934 |
| pH 1 0.5 at room temperature |
4.1 |
4.1 |
4.0 |
4.0 |
4.7 |
3.9 |
3.9 |
3.9 |
4.2 |
4.7 |