| Component (mg/L) |
M 6899 |
M 5519 |
M 0404 |
M 9274 |
M 7024 |
| Aluminum chloride • 6H2O |
|
| Ammonium nitrate |
1650.0 |
1650.0 |
1650.0 |
1650.0 |
1650.0 |
| Ammonium phosphate monobasic |
|
| Ammonium sulfate |
|
| Boric acid |
6.2 |
6.2 |
6.2 |
6.2 |
6.2 |
| Calcium chloride anhydrous |
332.2 |
332.2 |
332.2 |
332.2 |
332.2 |
| Calcium nitrate |
|
| Calcium phosphate tribasic |
|
| Cobalt chloride • 6H2O |
0.025 |
0.025 |
0.025 |
0.025 |
0.025 |
| Cupric sulfate • 5H2O |
0.025 |
0.025 |
0.025 |
0.025 |
0.025 |
| Na2-EDTA |
37.26 |
37.26 |
37.26 |
37.26 |
37.26 |
| ZnNa2-EDTA |
|
| Ferric chloride • 6H2O |
|
| Ferric citrate |
|
| Ferric sulfate |
|
| Ferric tartrate • 2H2O |
|
| Ferrous sulfate • 7H2O |
27.8 |
27.8 |
27.8 |
27.8 |
27.8 |
| Magnesium sulfate |
180.7 |
180.7 |
180.7 |
180.7 |
180.7 |
| Manganese chloride • 4H2O |
|
| Manganese sulfate • H2O |
16.9 |
16.9 |
16.9 |
16.9 |
16.9 |
| Molybdenum trioxide |
|
| Molybdic acid (sodium salt) • 2H2O |
0.25 |
0.25 |
0.25 |
0.25 |
0.25 |
| Nickel chloride • 6H2O |
|
| Nickel sulfate • 6H2O |
|
| Potassium chloride |
|
| Potassium iodide |
0.83 |
0.83 |
0.83 |
0.83 |
0.83 |
| Potassium nitrate |
1900.0 |
1900.0 |
1900.0 |
1900.0 |
1900.0 |
| Potassium phosphate monobasic |
170.0 |
170.0 |
170.0 |
170.0 |
170.0 |
| Potassium sulfate |
|
| Sodium nitrate |
|
| Sodium phosphate monobasic |
|
148.0 |
| Sodium sulfate |
|
| Zinc nitrate • 6H2O |
|
| Zinc sulfate • 7H2O |
8.6 |
8.6 |
8.6 |
8.6 |
8.6 |
| Organics (mg/L) |
M 6899 |
M 5519 |
M 0404 |
M 9274 |
M 7024 |
| Activated charcoal |
|
| Adenine hemisulfate |
|
80.0 |
| Agar |
|
|
|
8000.0 |
|
| 6-Benzylaminopurine (BA) |
|
| D-Biotin |
|
6gamma-gamma- Dimethylallylaminopurine |
|
30.0 |
| Glycine (free base) |
|
2.0 |
|
2.0 |
|
| Indole-3-acetic acid |
|
0.3 |
| Indole-3-butyric acid |
|
| Kinetin |
|
| MES (free acid) |
|
| myo-Inositol |
100.0 |
100.0 |
100.0 |
100.0 |
100.0 |
| alpha-Naphthaleneacetic acid |
|
| Nictotinic acid (free acid) |
|
0.5 |
1.0 |
0.5 |
|
| Peptone |
|
| Pyridoxine • HCl |
|
0.5 |
1.0 |
0.5 |
|
| Sucrose |
|
|
|
30,000 |
30,000 |
| Thiamine • HCl |
0.4 |
0.1 |
10.0 |
0.1 |
0.4 |
| |
M 6899 |
M 5519 |
M 0404 |
M 9274 |
M 7024 |
| Grams of powder to prepare 1 L |
4.4 |
4.4 |
4.4 |
42.4 |
34.7 |
| pH 1 0.5 at room temperature |
3.9 |
3.9 |
3.8 |
5.0 |
3.9 |