| Component (mg/L) |
K 4003 |
K 4128 |
P 0931 |
P 1056 |
P 6668 |
P 6793 |
M 5524* |
| Aluminum chloride • 6H2O |
|
| Ammonium nitrate |
|
500.0 |
825.0 |
825.0 |
825.0 |
825.0 |
1650.0 |
| Ammonium phosphate monobasic |
|
| Ammonium sulfate |
500.0 |
500.0 |
|
| Boric acid |
0.056 |
|
3.1 |
3.1 |
3.1 |
3.1 |
6.2 |
| Calcium chloride anhydrous |
|
|
166.0 |
166.0 |
166.0 |
166.0 |
332.2 |
| Calcium nitrate |
694.4 |
347.2 |
|
| Calcium phosphate tribasic |
|
| Cobalt chloride • 6H2O |
|
|
0.0125 |
0.0125 |
0.0125 |
0.0125 |
0.025 |
| Cupric sulfate • 5H2O |
0.0624 |
|
0.0125 |
0.0125 |
0.0125 |
0.0125 |
0.025 |
| Na2-EDTA |
|
|
37.24 |
37.24 |
37.24 |
37.24 |
37.26 |
| ZnNa2-EDTA |
|
| Ferric chloride • 6H2O |
|
| Ferric citrate |
|
| Ferric sulfate |
|
| Ferric tartrate • 2H2O |
|
| Ferrous sulfate • 7H2O |
25.0 |
25.0 |
27.85 |
27.85 |
27.85 |
27.85 |
27.8 |
| Magnesium sulfate |
122.125 |
122.125 |
90.35 |
90.35 |
90.35 |
90.35 |
180.7 |
| Manganese chloride • 4H2O |
|
| Manganese sulfate • H2O |
5.682 |
5.682 |
8.45 |
8.45 |
8.45 |
8.45 |
16.9 |
| Molybdenum trioxide |
0.016 |
|
| Molybdic acid (sodium salt) • 2H2O |
|
|
0.125 |
0.125 |
0.125 |
0.125 |
0.25 |
| Nickel chloride • 6H2O |
|
| Nickel sulfate • 6H2O |
|
| Potassium chloride |
|
250.0 |
|
| Potassium iodide |
|
|
0.415 |
0.415 |
0.415 |
0.415 |
0.83 |
| Potassium nitrate |
|
|
950.0 |
950.0 |
950.0 |
950.0 |
1900.0 |
| Potassium phosphate monobasic |
250.0 |
250.0 |
85.0 |
85.0 |
85.0 |
85.0 |
170.0 |
| Potassium sulfate |
|
| Sodium nitrate |
|
| Sodium phosphate monobasic |
|
| Sodium sulfate |
|
| Zinc nitrate • 6H2O |
|
| Zinc sulfate • 7H2O |
0.331 |
|
5.3 |
5.3 |
5.3 |
5.3 |
8.6 |
| Organics (mg/L) |
K 4003 |
K 4128 |
P 0931 |
P 1056 |
P 6668 |
P 6793 |
M 5524* |
| Activated charcoal |
|
|
|
2000.0 |
2000.0 |
|
|
| Adenine hemisulfate |
|
| Agar |
|
| Banana powder (50% malto-dextrin) |
|
|
|
30,000 |
|
|
|
| 6-Benzylaminopurine (BA) |
|
2.0 |
|
| D-Biotin |
|
6gamma-gamma- Dimethylallylaminopurine |
|
| Glycine (free base) |
|
| Indole-3-acetic acid |
|
| Indole-3-butyric acid |
|
| Kinetin |
|
| MES (free acid) |
|
|
1000.0 |
1000.0 |
1000.0 |
1000.0 |
|
| myo-Inositol |
|
|
100.0 |
100.0 |
100.0 |
100.0 |
|
| alpha-Naphthaleneacetic acid |
|
0.5 |
|
| Nictotinic acid (free acid) |
|
|
1.0 |
1.0 |
1.0 |
0.5 |
|
| Peptone |
|
|
2000.0 |
2000.0 |
2000.0 |
2000.0 |
|
| Pyridoxine • HCl |
|
|
1.0 |
1.0 |
1.0 |
0.5 |
|
| Sucrose |
20,000 |
20,000 |
20,000 |
20,000 |
20,000 |
20,000 |
|
| Thiamine • HCl |
|
|
10.0 |
10.0 |
10.0 |
1.0 |
|
| |
K 4003 |
K 4128 |
P 0931 |
P 1056 |
P 6668 |
P 6793 |
M 5524* |
| Grams of powder to prepare 1 L |
21.6 |
22.0 |
25.3 |
57.3 |
27.3 |
25.3 |
4.3 |
| pH ± 0.5 at room temperature |
4.7 |
4.8 |
5.0 |
5.2 |
5.4 |
5.0 |
3.9 |
|
*Composition for M 5524, Murashige and Skoog Basal Salt Mixture, listed for purposes of comparison only.