Sigma-Aldrich
Life Science > Molecular Biology > Plant Biotechnology > Tissue Culture Protocols > Silver Thiosulfate
Plant Tissue Culture Protocols
Silver Thiosulfate
    Prepare a 0.1 M Sodium Thiosulfate (STS) stock solution by dissolving 1.58 g of Sodium Thiosulfate (Product No. S 7026) into 100 ml of water. Prepare a 0.1 M Silver Nitrate stock solution by dissolving 1.7 g of Silver Nitrate (Product No. S 7276) into 100 ml of water. Store the stock solutions in the dark until needed to prepare the STS.
    In general, the STS is prepared with a molar ratio between silver and thiosulfate of 1:4, respectively. Nearly all of the silver present in the solution is in the form of
[Ag(S2O3)2]3-, the active complex for ethylene effect inhibition.
    Prepare a 0.02 M STS by slowly pouring 20 ml of 0.1 M silver nitrate stock solution into 80 ml of 0.1 M sodium thiosulfate stock solution. The STS can be stored in the refrigerator for up to a month. However, preparation of the STS just prior to use is recommended.