|
Whole Genome Amplification
Buccal Swab
|
This protocol provides a simple and convenient method to isolate, amplify, and purify genomic DNA from buccal swabs. Buccal swabs are a convenient method of acquiring a DNA sample. Once the DNA is isolated using the following extraction protocol, it can be amplified using the GenomePlex® Whole Genome Amplification Protocol to provide enough DNA for numerous downstream applications.
Required Products
- GenElute™ Mammalian Genomic Miniprep Kit (Product No. G1N10)
Materials to be Supplied by the User
- Buccal swab
- 1.5 ml microcentrifuge tubes
- Ethanol (Product No. E7023)
- Microcentrifuge (with rotor for 2 ml tubes)
- Water, molecular biology reagent (Product No. W4502)
- 55 °C water bath or heat block
Extraction of DNA from Buccal Swab It is recommended to use the GenElute Mammalian Genomic DNA Miniprep Kit (Product No. G1N10) for this process.
- Dry the collected swabs at room temperature for 15 minutes.
- Add 280 µl of Lysis Solution T and 20 µl of Proteinase K. Insert the swab and gently spin. Cap the tube and mix by vortexing.
- Incubate the sample at 55 °C for 20 minutes with occasional vortexing.
- Add 200 µl of Lysis Solution C and vortex thoroughly for 15 seconds.
- Incubate at 70 °C for 10 minutes.
- Add 500 µl of Column Preparation Solution to each GenElute Miniprep Binding Column (red
O-ring) and centrifuge at 12,000 × g for 1 minute. Discard the flow-though liquid. Note: The Column Preparation Solution maximizes binding of DNA to the membrane resulting in more consistent yields.
- Add 200 µl of 95–100% ethanol to the lysate from step 5.
- Mix thoroughly by vortexing and add the entire contents of the tube into the binding column.
- Centrifuge at ≥6500 × g for 1 minute.
- Discard the collection tube containing the flow-through and place the binding column in a new 2 ml collection tube.
- Add 500 µl of Wash Solution (be sure to dilute with ethanol prior to first use) and centrifuge at
≥6500 × g for 1 minute.
- Discard the collection tube and flow-through and place the binding column in a new 2 ml collection tube.
- Add another 500 µl of Wash Solution to the binding column and centrifuge at maximum speed (12,000–16,000 × g) for 3 minutes to dry the binding column.
- Pipette 200 µl of Elution Solution onto the binding column and centrifuge for 1 minute at
≥6500 × g.
- Store the eluted DNA at –20 °C or proceed to the amplification step.
Application Data
GenomePlex Whole Genome Amplification Performed on a Buccal Swab Sample
|
|
|
Legend: Amplified Sigma products are visualized on 1.5% agarose gel. 5 µl of amplified product was loaded per well. The GenomePlex amplified products result in an average size of 400 bp . The smear pattern varies by source as shown on the gel.
|
|
Lane 1—1 kb Ladder Lane 2—Blood Lane 3—Plant Lane 4—Buccal Swab
|
Lane 5—Soil Lane 6—Positive Control Lane 7—1 kb Ladder