Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
PCR
Genotyping Data
Accurate Amplification Preserving Genome Representation
Single Cell WGA (WGA4) Preserves Genotype of the Original Genome.
Single Cell Whole Genome Amplification Genotyping Data Female sample
Female sample
Positive control
Negative control
Single Cell WGA was performed on amniotic fluid sourced fibroblast cells drawn from two different patients. The genotyping control samples provide no signals in the negative control and male specific peaks for the positive control. The two upper graphs represent data for two different female samples as amplified with the single cell WGA kit. These data indicate that GenomePlex single cell amplified products are compatible with gender study assays.
back to Single Cell Whole Genome Amplification Kit