Sigma-Aldrich
Life Science > Molecular Biology > Whole Genome Amplification > Single Cell Kits > Short Tandem Repeat Analysis
Whole Genome Amplification
Short Tandem Repeat Analysis
Single Cell WGA (WGA4) Supports Robust Amplification
Single Cell Whole Genome Amplification STR Data
Lane 1: Marker
Lane 2: Positive Control
Lane 3: Negative Control
Lane 4: cell 1
Lane 5: cell 2
Lane 6: cell 3
Lane 7: cell 4
Lane 8: cell 5
Five fresh lymphocytes were lysed and amplified using the Single Cell WGA Kit (WGA4). The measure of success was performed with the DXS1623 microsatellite marker, amplified by two-rounds PCR. The final product was verified on a 1.5% agarose gel. No contamination was noted in the negative control sample.
Data is courtesy of Dr. Christophe Beroud from Laboratoire de Genetique Moleculaire et Chromosomique, Institute Universitaire de Recherche Clinique, France.
back to Single Cell Whole Genome Amplification Kit