Sigma-Aldrich
Life Science > Molecular Biology > Whole Genome Amplification > WGA Tissue Kit
Whole Genome Amplification
WGA Tissue Kit
Too many samples? Now available: WGA and WTA Services
Advances in Genetic Analysis of Archived Samples
Advances in Genetic Analysis of Archived Samples
On-Demand Webcast
The GenomePlex Tissue Whole Genome Amplification Kit (WGA5) contains all of the reagents necessary to amplify genomic DNA directly from formalin-fixed, paraffin-embedded (FFPE), frozen, RNAlater®-preserved or fresh tissue. The kit includes optimized reagents for tissue disruption and cell lysis eliminating the need for tedious organic extractions to remove excess paraffin or DNA purification prior to amplification. This rapid and straightforward method yields microgram quantities of genomic DNA from one milligram of tissue.
DNA isolated from archived tissue is often of poor quality making it unsuitable for most commercially available whole genome amplification kits that require high quality DNA as starting material. The flexibility of the GenomePlex technology enables the use of compromised starting material, such as DNA isolated from FFPE tissue, for whole genome amplification.
Features
  • Streamlined protocol
    Optimized extraction protocol: no deparaffinization or DNA purification necessary
  • Robust and Accurate Amplification
    Suitable for direct amplification from fresh or frozen tissue, FFPE or RNAlater® -preserved samples with no detectable allele bias
  • Unlimited Genetic Analysis
    Identification of genome-wide copy number and chromosomal abnormalities from archived tissue
back to top
Ordering Information
Product Code Product Name Technical Bulletin
WGA5 GenomePlex® Tissue Whole Genome Amplification (WGA) Kit sg_dd_hts_tbicon
GenomePlex and OmniPlex are Registered Trademarks of Rubicon Genomics. This product is for research use only. Use of this product for human or animal therapeutic or diagnostic purposes requires licensing from Rubicon Genomics, Inc. Unauthorized use violates proprietary rights of Rubicon Genomics, Inc (www.rubicongenomics.com).
TaqMan® is a registered trademark of Roche Molecular Systems, Inc.
back to top
back to Whole Genome Amplification