Sigma-Aldrich
Life Science > Molecular Biology > Whole Genome Amplification > Whole Transcriptome Amplification
Whole Genome Amplification
Whole Transcriptome Amplification
Too many samples? Now available: WGA and WTA Services
Rapid Amplification of Total RNA
Amplify RNA, WTA2-TransPlex, Whole Transcriptome AmplificationThe TransPlex Whole Transcriptome Amplification (WTA1 and WTA2) Kits provide an accurate, fast and simple method of amplifying total RNA from a variety of sources including blood, fixed and frozen tissue, cell culture, FACS sorted cells, plants and microorganisms. The TransPlex WTA technology accurately amplifies total RNA by the utilization of a unique blend of quasi-random primers to ensure accurate transcriptome coverage and rapid amplification of total RNA. The resulting cDNA is suitable for qPCR, micro array, and traditional cloning.
  • Product Description
  • NEW! Labeling Protocols for Microarrays
  • FAQs
  • Posters and Publications
Product Description
The TransPlex WTA kits provide rapid amplification of total RNA in less than 4 hours without 3’-bias. Amplified RNA (cDNA) is suitable for qPCR, micro array analysis, and traditional cloning. The TransPlex WTA process has two steps; 1) library synthesis and 2) library amplification. To synthesize the cDNA library, sample RNA is incubated with reverse transcriptase and non-self-complementary primers comprised of a quasi-random 3’ end and a universal 5’ end. Annealed primers are extended by WTA polymerase, and displaced single strands become new templates for primer annealing and extension. The process creates an OmniPlex cDNA library comprised of random, overlapping 100-1000 base fragments, flanked by a universal end sequence. Finally, universal-primer PCR amplifies the OmniPlex® cDNA library to produce microgram quantities of WTA products.
Product Number Product Description Reactions
WTA1-10RXN TransPlex Whole Transcriptome Amplification (WTA) Kit 10 reactions
WTA1-50RXN TransPlex Whole Transcriptome Amplification (WTA) Kit 50 reactions
WTA2-10RXN Complete TransPlex Whole Transcriptome Amplification (WTA) Kit 10 reactions
WTA2-50RXN Complete TransPlex Whole Transcriptome Amplification (WTA) Kit 50 reactions
NEW! Labeling Protocols for Microarrays
  • Affymetrix WTA Application Note (378 Kb PDF)
  • Agilent WTA Application Note (840 Kb PDF)
  • NimbleGen WTA Application Note (435 Kb PDF)
  • Illumina WGA Application Note (202 Kb PDF)
FAQs
TransPlex Whole Transcriptome Amplification Kit FAQs
Posters and Publications
Modification of the Transplex™ WTA2 Amplification Product for Next Generation Sequencing (1.61 Mb PDF)
Contact techserv@sial.com to participate as an early adopter of the WTA2-Sequencing technology.
Accurate Expression Profiling of Very Small Cell Populations (520 Kb PDF)
Virus-Plus-Susceptibility Gene Interaction Determines Crohn's Disease Gene Atg16L1 Phenotypes in Intestine (1.2 Mb PDF)
TransPlex WTA2 on FFPE samples a comparison with NuGen (2.99 Mb PDF)
TransPlex WTA2 amplification of very low RNA input and single cells, using qPCR analysis (1.25 Mb PDF)
Expression Profiling of Fixed and Unfixed Tissue Using Transplex™ Whole Transcriptome Amplification Kits (140 Kb PDF)
Whole Transcriptome Amplification from Degraded RNA Transcripts (1.09 Mb PDF)
FACS Sorting of Stem Cells and Whole Transcriptome Amplification (111 Kb PDF)
back to top
back to Whole Genome Amplification